The total volume in this item is 25 mL.
Recommended Products
biological source
bovine pancreas
Quality Level
form
liquid
specific activity
1500 units/mL
3000 units/mL
concentration
20 mg/mL in Tris buffer
technique(s)
DNA purification: suitable
storage temp.
room temp
Gene Information
bovine ... RNASE1(282340) , RNASE1(280930)
General description
For sample preparation in GenElute™ kits.
Application
Biochem/physiol Actions
Preparation Note
Legal Information
sample prep
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
what is the total volume of this product?
1 answer-
Helpful?
-
-
What is the concentration of the RNase a solution in product R6148?
1 answer-
R6148 is solution of 20 mg/ml RNAse A in Tris Buffer with 50% glycerol. A rough estimate of activity is 1000-2000 Kunitz units/ml.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service