Skip to Content
Merck
All Photos(1)

Key Documents

NA0150

Sigma-Aldrich

GenElute HP Plasmid Miniprep Kit

greener alternative

sufficient for 70 preparations

Synonym(s):

GenElute HP Plasmid Kit, HP miniprep kit, fast miniprep kit, fast plasmid isolation kit, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, ultrafast plasmid isolation kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 70 preparations

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA extraction: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are then removed by a vacuum or spin wash steps. Finally, the bound plasmid DNA is eluted in water or Tris-EDTA buffer.

The GenElute HP Plasmid Miniprep Kit offers an ultrafast and efficient solution for plasmid preparation from E. coli cultures. This kit combines silica-based membrane technology and the convenience of spin or vacuum format,up to 25 μg of high copy plasmid DNA can be recovered from 1-5 mL of E. coli culture in less than 30 minutes. Note that actual yield and optimum volume of culture to use depend on the plasmid and the culture medium.
The GenElute HP Plasmid Miniprep Kit offers an ultrafast and efficient solution for plasmid preparation from E. coli cultures. This kit combines silica-based membrane technology and the convenience of spin or vacuum format. An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are then removed by a vacuum or spin wash steps. Finally, the bound plasmid DNA is eluted in water or Tris-EDTA buffer.

The recovered plasmid DNA is ready for immediate use in applications such as restriction digest, cloning, PCR, transfection, and sequencing.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The recovered plasmid DNA is ready for immediate use in applications such as restriction enzyme digestion, cloning, PCR, transcription, and sequencing.

Features and Benefits

  • Purified plasmid DNA in less than 30 minutes
  • Up to 25 μg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • No phenol/chloroform extraction or alcohol precipitation required

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine Guild et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 67(Pt 9), 1027-1031 (2011-09-10)
Recombinant expression of proteins of interest in Escherichia coli is an important tool in the determination of protein structure. However, lack of expression and insolubility remain significant challenges to the expression and crystallization of these proteins. The SSGCID program uses
Antonietta Carbone et al.
Acta biochimica Polonica, 59(2), 275-278 (2012-06-14)
Supercoiled state corresponds to the active form for plasmid applications. The relaxed circular form of plasmids is often inactive or poorly active. To obtain significant amounts of almost fully supercoiled DNA, we modified the standard protocol of a commercially available
Radhika Goenka et al.
The Journal of experimental medicine, 211(1), 45-56 (2013-12-25)
We have assessed the role of B lymphocyte stimulator (BLyS) and its receptors in the germinal center (GC) reaction and affinity maturation. Despite ample BLyS retention on B cells in follicular (FO) regions, the GC microenvironment lacks substantial BLyS. This
María Pilar Trotonda et al.
Journal of bacteriology, 187(16), 5790-5798 (2005-08-04)
The biofilm-associated protein Bap is a staphylococcal surface protein involved in biofilm formation. We investigated the influence of the global regulatory locus sarA on bap expression and Bap-dependent biofilm formation in three unrelated Staphylococcus aureus strains. The results showed that
S Weidlich et al.
British journal of cancer, 105(2), 246-254 (2011-06-30)
The epidermal growth factor receptor-targeted monoclonal antibody cetuximab (Erbitux) was recently introduced for the treatment of metastatic colorectal cancer. Treatment response is dependent on Kirsten-Ras (K-Ras) mutation status, in which the majority of patients with tumour-specific K-Ras mutations fail to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service