Skip to Content
Merck
All Photos(3)

Documents

R0404

Sigma-Aldrich

Ramipril

≥98% (HPLC)

Synonym(s):

Tritace, [2S,3aS,6aS]-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N2O5
CAS Number:
Molecular Weight:
416.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ~18 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

O=C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O)=O)[C@H](C)N[C@H](C(OCC)=O)CCC3=CC=CC=C3

InChI

1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18-,19-,20-/m0/s1

InChI key

HDACQVRGBOVJII-JBDAPHQKSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Zoja et al.
American journal of physiology. Renal physiology, 304(6), F808-F819 (2012-11-09)
Bardoxolone methyl is an antioxidant inflammation modulator acting through induction of Keap1-Nrf2 pathway. Results from a recent phase IIb clinical trial reported that bardoxolone methyl was associated with improvement in the estimated glomerular filtration rate in patients with advanced chronic
Daniela Dunkler et al.
JAMA internal medicine, 173(18), 1682-1692 (2013-08-14)
Type 2 diabetes mellitus and associated chronic kidney disease (CKD) have become major public health problems. Little is known about the influence of diet on the incidence or progression of CKD among individuals with type 2 diabetes. To examine the
Guillaume Pare et al.
Pharmacogenetics and genomics, 23(9), 470-478 (2013-07-11)
The objective of this study was to identify genetic variants associated with angiotensin-converting enzyme (ACE) inhibitor-associated angioedema. We carried out a genome-wide association study in 175 individuals with ACE inhibitor-associated angioedema and 489 ACE inhibitor-exposed controls from Nashville (Tennessee) and
G Castoldi et al.
American journal of nephrology, 37(1), 65-73 (2013-01-19)
Diabetic nephropathy is the main cause of end-stage renal disease. N-acetyl-seryl-aspartyl-lysyl-proline (Ac-SDKP), a physiological tetrapeptide hydrolyzed by the angiotensin-converting enzyme (ACE), has antifibrotic effects in the cardiovascular system and in the kidney in experimental models of hypertension, heart failure and
Joshua I Barzilay et al.
Clinical journal of the American Society of Nephrology : CJASN, 8(2), 233-240 (2012-11-28)
The microvascular circulation plays an important role in bone health. This study examines whether albuminuria, a marker of renal microvascular disease, is associated with incident hip and pelvic fractures. This study reanalyzed data from the Ongoing Telmisartan Alone and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service