Skip to Content
Merck

Skip To

SML0101

Montelukast sodium hydrate

≥98% (HPLC), CysLT1 receptor antagonist, powder

Synonym(s):

1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic acid sodium salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C35H35ClNO3S·Na · xH2O
Molecular Weight:
608.17 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Montelukast sodium hydrate, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +90 to +106° in methanol (c=1)

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥8 mg/mL at 60 °C

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

O.[Na+].CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC2)CC([O-])=O)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3

InChI

1S/C35H36ClNO3S.Na.H2O/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;;/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39);;1H2/q;+1;/p-1/b15-10+;;/t32-;;/m1../s1

InChI key

QAXHQHBRRJHZHL-ZPQUALOVSA-M

Application

Montelukast sodium hydrate has been used as a positive control drug to study the protective effects of Gumiganghwal-tang aqueous extract (GGTA) against airway inflammation and pulmonary fibrosis.

Biochem/physiol Actions

Montelukast sodium hydrate is a leukotriene receptor antagonist (LTRA) used for the maintenance treatment of asthma and to relieve symptoms of seasonal allergies. It is a subtype specific CysLT1 receptor antagonist.
Montelukast sodium hydrate is subtype specific CysLT1 receptor antagonist; Antiasthmatic; antiallergic

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Leukotriene Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0038SML0204S8825
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: ≥8 mg/mL at 60 °C

solubility

H2O: ≥9 mg/mL

solubility

DMSO: ≥10 mg/mL at 60 °C

solubility

DMSO: >10 mg/mL

storage condition

desiccated

storage condition

desiccated

storage condition

-

storage condition

desiccated


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters