Skip to Content
Merck
All Photos(1)

Documents

PZ0106

Sigma-Aldrich

SC-236

≥98% (HPLC)

Synonym(s):

4-[5-(4-Chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide, SC-58236

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11ClF3N3O2S
CAS Number:
Molecular Weight:
401.79
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

NS(=O)(=O)c1ccc(cc1)-n2nc(cc2-c3ccc(Cl)cc3)C(F)(F)F

InChI

1S/C16H11ClF3N3O2S/c17-11-3-1-10(2-4-11)14-9-15(16(18,19)20)22-23(14)12-5-7-13(8-6-12)26(21,24)25/h1-9H,(H2,21,24,25)

InChI key

NSQNZEUFHPTJME-UHFFFAOYSA-N

Application

SC-236 has been used as a COX-2 inhibitor to study its effects on the mechano-reflex in rats.

Biochem/physiol Actions

SC-236 exhibits anti-tumor activity in gastric cancer cells by modulating activator protein-1 (AP-1) expression and by blocking the anchorage-independent cell growth. It also exhibits a protective effect against cartilage damage by minimizing the inflammation and pain in osteoarthritis. It is also reported to treat allergic inflammation.
SC-236 is a cyclooxygenase-2 (COX-2) inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Meurer et al.
Pflugers Archiv : European journal of physiology, 470(11), 1691-1703 (2018-07-22)
Endotoxemia-related acute kidney injury (AKI) is associated with increased formation of prostaglandins, which may serve as a compensatory mechanism to maintain renal function. We hypothesized that an increase of renal EP2 or EP4 receptors and/or a downregulation of renal EP1
Hae-Kyoung Kim et al.
International archives of allergy and immunology, 171(1), 61-70 (2016-11-14)
Cytosolic phospholipase A2 (cPLA2) plays a key role in the development of late-phase anaphylaxis. L-Glutamine (Gln), a nonessential amino acid, has anti-inflammatory activity via inhibiting cPLA2. We used a penicillin-induced murine model of anaphylaxis, and late-phase anaphylaxis was quantified by
Ariel Morales et al.
Experimental physiology, 97(8), 943-954 (2012-04-24)
Cyclo-oxygenase (COX) enzymes are responsible for the formation from arachidonic acid of prostaglandins, among other metabolites. Prior studies have suggested that inhibition of the COX pathway attenuates the responses of sympathetic nerve activity and blood pressure during static muscle contraction.
Benjamin Chun-Yu Wong et al.
Gastroenterology, 126(1), 136-147 (2003-12-31)
Aspirin exerts antitumor effect partly through blocking tumor promoter-induced activator protein-1 (AP-1) activation. The aim of this study is to determine how specific COX-2 inhibitor SC-236 mediates antitumor effect by modulation of AP-1-signaling pathway. AP-1 transcriptional activity and DNA-binding activity
Patrick Slattery et al.
American journal of physiology. Renal physiology, 310(10), F1113-F1122 (2016-03-18)
Deletion of cyclooxygenase (COX)-2 causes impairment of kidney development, including hypothrophic glomeruli and cortical thinning. A critical role for COX-2 is seen 4-8 days postnatally. The present study was aimed at answering whether different COX-2 gene dosage and partial pharmacological

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service