Skip to Content
Merck

Skip To

M9766

4-Methylumbelliferyl α-D-glucopyranoside

α-glucosidase substrate, fluorogenic, ≥99% (TLC), powder

Synonym(s):

4-Methylumbelliferyl α-D-glucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
241-794-0
MDL number:
Beilstein/REAXYS Number:
1690776
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

4-Methylumbelliferyl α-D-glucopyranoside, α-glucosidase substrate

Quality Segment

assay

≥99% (TLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)ccc12

InChI

1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15-,16+/m1/s1

InChI key

YUDPTGPSBJVHCN-JZYAIQKZSA-N

Application

4-Methylumbelliferyl ǥ-D-glucopyranoside has been used to assay acid alpha-glucosidase (GAA) activity in tissue homogenates.[1][1][2]

Biochem/physiol Actions

4-Methylumbelliferyl ǥ-D-glucopyranoside serves as a fluorogenic substrate for the ǥ-glucosidase enzyme. The product, 4-methylumbelliferyl, shows a peak at 440nm in the fluorescence spectra.[3]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M0905M9659M7008
assay

≥99% (TLC)

assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

solubility

pyridine: 10 mg/mL, clear, colorless to faintly yellow

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
M9766-10MG04061834064565
M9766-25MG04061835557783
M9766-100MG04061834064558
M9766-250MG04061835546190
M9766-1G04061835557776

Questions

Reviews

No rating value

Active Filters