Skip to Content
Merck
All Photos(1)

Key Documents

PZ0004

Sigma-Aldrich

Varenicline tartrate

≥98% (HPLC)

Synonym(s):

7,8,9,10-Tetrahydro-6,10-methano-6H-pyrazino[2,3-h][3]benzazepine tartrate, Champix

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3 · C4H6O6
CAS Number:
Molecular Weight:
361.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

room temp

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.C1NC[C@H]2C[C@@H]1c3cc4nccnc4cc23

InChI

1S/C13H13N3.C4H6O6/c1-2-16-13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15-1;5-1(3(7)8)2(6)4(9)10/h1-2,4-5,8-9,14H,3,6-7H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-,9+;1-,2-/m.1/s1

InChI key

TWYFGYXQSYOKLK-CYUSMAIQSA-N

Application

Varenicline tartrate has been used to investigate its effect on sensorimotor gating in rats under prepulse inhibition (PPI). It is suitable for use as a reference standard in capillary isotachophoresis (ITP) coupled on-line with capillary zone electrophoresis (CZE) for the quantification of varenicline metabolite in urine samples. It is also suitable for use for testing its effect on locomotor and nonlocomotor behavioral deficits in rats.

Biochem/physiol Actions

Varenicline tartrate is a partial α4β2 nicotinic receptor agonist and α7 full agonist. Varenicline competitively binds to α4β2 receptors and partially stimulates without creating a full nicotine effect, while simultaneoudly blocking the ability of nicotine to bind to the receptors. Varenicline thus blocks the ability of nicotine to activate α4β2 receptors and stimulate the central nervous mesolimbic dopamine system, believed to be the neuronal mechanism underlying reinforcement and reward experienced upon smoking.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of varenicline on behavioral deficits in a rat model of Parkinson?s disease induced by unilateral 6-hydroxydopamine lesion of substantia nigra
Tan R, et al.
Behavioural Pharmacology, 29(4), 327-335 (2018)
Helen M Kamens et al.
Brain research bulletin, 138, 20-25 (2017-08-06)
Adolescence is a critical period in brain development that coincides with the initiation of alcohol use. Nicotinic acetylcholine receptors (nAChR) have been shown to modulate ethanol behaviors in adult humans and in animal models; however, the role of these receptors
Rachel L Schassburger et al.
Psychopharmacology, 232(5), 975-983 (2014-09-12)
Varenicline (VAR), a smoking cessation aid that is a partial agonist at nicotinic receptors, mimics the reinforcement-enhancing effects of nicotine. Varenicline, when accompanied by non-drug cues, is self-administered by rats, though it is unclear whether this results from varenicline acting
Kelly J Clemens et al.
Addiction biology, 22(2), 400-410 (2015-12-03)
An array of pharmacological and environmental factors influence the development and maintenance of tobacco addiction. The nature of these influences likely changes across the course of an extended smoking history, during which time drug seeking can become involuntary and uncontrolled.
Elizabeth G Guy et al.
Behavioural pharmacology, 25(8), 775-783 (2014-09-18)
Smoking tobacco remains one of the leading causes of preventable deaths in North America. Nicotine reinforces smoking behavior, in part, by enhancing the reinforcing properties of reward-related stimuli, or conditioned stimuli (CSs), associated with tobacco intake. To investigate how pharmaceutical

Articles

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service