Skip to Content
Merck
All Photos(1)

Documents

712264

Sigma-Aldrich

Cytisine

≥99.0% (HPLC), ≥99%

Synonym(s):

(−)-Cytisine, (1R,5S)-1,2,3,4,5,6-Hexahydro-1,5-methano-8H-pyrido[1,2a][1,5]diazocin-8-one, (1S,9S)-3,11-Diazatricyclo[7.3.1.03,8]trideca-5,7-dien-4-one, Baptitoxin, Laburnin, Sophorine, Ulexine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%
≥99.0% (HPLC)

form

powder

optical activity

[α]/D -108±3°, c = 1% in ethanol

bp

218 °C/2 mmHg (lit.)

mp

154-156 °C (lit.)

SMILES string

O=C1C=CC=C2C3CNCC(C3)CN12

InChI

1S/C11H14N2O/c14-11-3-1-2-10-9-4-8(5-12-6-9)7-13(10)11/h1-3,8-9,12H,4-7H2/t8-,9+/m0/s1

InChI key

ANJTVLIZGCUXLD-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cystine ((1R,5S)-1,2,3,4,5,6-hexahydro-1,5-methano-8H-pyrido[1,2a][1,5]diazocin-8-one) displays classical antidepressant action in various animal models of antidepressant efficacy. Infrared and Raman spectral studies suggest that it has two stable conformers.
Cytisine was found to be more effective than nicotine-replacement therapy in helping smokers to quit smoking. It is an α4 β2 nicotinic receptor partial agonist. It is a plant alkaloid which is commercially extracted for use as a smoking cessation medication. Preparation of nicotine partial agonist cytisine via “in situ” Stille or Suzuki biaryl pyridine coupling has been reported. Cytisine is a competitive partial agonist at α4 subunit-containing nicotinic acetylcholine (nACh) receptors while at homomeric α7-nACh receptors it behaves as a full agonist with a relatively lower potency.

Application

Cystine may be used as nicotine substitute in nicotine addiction therapy.
Cytisine is suitable for use in a study to compare the effects of varenicline and cytisine on ethanol consumption by rats bred for many generations as high ethanol drinkers.

Biochem/physiol Actions

Potent agonist at α3β4 and α7 nicotinic acetylcholine receptors and partial agonist at α4β2 nicotinic acetylcholine receptors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Experimental (FT-IR and Raman) and theoretical (DFT) studies on the vibrational dynamics in cytisine.
Gornicka E, et al.
Vibrational Spectroscopy, 36(1), 105-115 (2004)
Y E Slater et al.
Neuropharmacology, 44(4), 503-515 (2003-03-21)
Cytisine (cy) is a potent and competitive partial agonist at alpha4 subunit-containing nicotinic acetylcholine (nACh) receptors while at homomeric alpha7-nACh receptors it behaves as a full agonist with a relatively lower potency. In the present study, we assessed the effects
Natalie Walker et al.
BMC public health, 11, 880-880 (2011-11-23)
Smokers need effective support to maximise the chances of successful quit attempts. Current smoking cessation medications, such as nicotine replacement therapy (NRT), bupropion, nortriptyline or varenicline, have been shown to be effective in clinical trials but are underused by smokers
Michael J Marks et al.
Journal of neurochemistry, 130(2), 185-198 (2014-03-26)
Nicotinic acetylcholine receptors (nAChR) of the α6β2* subtype (where *indicates the possible presence of additional subunits) are prominently expressed on dopaminergic neurons. Because of this, their role in tobacco use and nicotine dependence has received much attention. Previous studies have
Edwin G Pérez et al.
Natural product reports, 29(5), 555-567 (2012-03-01)
Covering: up to the end of 2011. This review covers classical and modern structural modifications of the alkaloid, the more recent (since 2007) syntheses of cytisine and analogues, and the pharmacology of these compounds, with emphasis on their interactions with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service