Skip to Content
Merck
All Photos(2)

Key Documents

P7798

Sigma-Aldrich

Polysucrose 400

powder

Synonym(s):

Sucrose-epichlorohydrin copolymers

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

100
200

form

powder

mol wt

300,000-550,000

color

white

solubility

water: soluble

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Polysucrose is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relates to the size of many normal food proteins.

Application

Polysucrose has been used in a study to develop a new method for the purification of human motile spermatozoa. It has also been used in a study that investigated the antiproliferative and antitumor activities of D-reverse peptides.
Used for cell separation and organelle isolation.

Other Notes

A nonionic synthetic polymer of sucrose.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Linkage

Similar to Ficoll 400, but from a different supplier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N H Guo et al.
The journal of peptide research : official journal of the American Peptide Society, 50(3), 210-221 (1997-10-06)
The extracellular matrix glycoprotein thrombospondin-1 (TSP1) inhibits angiogenesis, endothelial cell growth, motility and adhesion. Peptides from the type I repeats of TSP1 mimic the adhesive and growth inhibitory activities of the intact protein and specifically interact with heparin and transforming
C Y Andersen et al.
Journal of assisted reproduction and genetics, 14(10), 624-628 (1998-02-03)
A newly developed method for the isolation of human motile spermatozoa using density-gradient centrifugation was compared with the traditionally used Percoll technique. Sperm samples were divided into two equal aliquots, which were purified with either the traditionally performed Percoll technique
H Oman et al.
International archives of allergy and immunology, 98(3), 220-226 (1992-01-01)
To measure intestinal absorption of macromolecules we have developed a new technique employing synthetic polysucrose polymers as probe molecules. Polysucrose (PS) is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relates

Articles

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service