Skip to Content
Merck

Skip To

F2637

Ficoll® 400

BioXtra, lyophilized powder

Synonym(s):

Polysucrose 400, Ficoll 400

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.52
MDL number:
eCl@ss:
32190102
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

grade

Molecular Biology

product line

BioXtra

form

lyophilized powder

foreign activity

DNase (Exonuclease), NICKase (Endonuclease), RNase and Protease, none detected

storage temp.

room temp

General description

Ficoll® 400 is a non-ionic synthetic polymer of sucrose used for cell separation and organ isolation.

Application

Suitable for
  • hybridization experiments to reduce non-specific binding of labeled probes to the membrane
  • preparation of DNA loading buffer (15% Ficoll®, 0.25% bromophenol blue and 0.25% xylene cyanol FF
  • separation of cells with similar density but different size (PBMC from human peripheral blood, bone marrow and umbilical cord blood)
  • producing density gradients to separate subcellular components
  • vitrification of cell and tissue samples.
  • to compare the effect of different perfusates on Hypoxic pulmonary vasoconstriction (HPV).
Used for cell separation and organelle isolation.

Features and Benefits

  • free from DNases, RNases, phosphatases and proteases
  • preserves functional and morphological integrity of the separated cells

Other Notes

A nonionic synthetic polymer of sucrose.

Legal Information

Ficoll is a registered trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F8016F9378F4375
description

BioXtra, lyophilized powder

description

lyophilized powder, Type 400-DL, BioReagent, suitable for cell culture

description

Type 400-DL, lyophilized powder

description

Type 400

grade

Molecular Biology

grade

-

grade

-

grade

-

biological source

non-animal source

biological source

-

biological source

sugar beets (Beta vulgaris)

biological source

non-animal source

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

-

form

lyophilized powder

form

powder, spray-dried

storage temp.

room temp

storage temp.

-

storage temp.

15-25°C

storage temp.

room temp

product line

BioXtra

product line

BioReagent

product line

-

product line

-


Still not finding the right product?

Explore all of our products under Ficoll® 400


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. Is Product F2637, Ficoll, the same as Ficoll 400?

    1 answer
    1. Yes. It is Ficoll 400.  This product is graded molecular biology grade and is free of any nucleases

      Helpful?

  2. Can you use Product F2637, Ficoll, to isolate mononuclear cells?

    1 answer
    1. When trying to isolate mononuclear cells you can use this product but we offer Histopaque that is a mixture of Ficoll and Hypaque. For humans, we offer H8889, Histopaque®-1077 Hybri-Max™ or 1077-1, Histopaque®-1077. For rodents we offer 1083-1, Histopaque®-1083. For isolation of neutrophils, we offer 1119-1, Histopaque®-1119.

      Helpful?

  3. Do you have a protocols for use of Product F2637, Ficoll, in various applications?

    1 answer
    1. Please check the product information sheet for F4375 which has a detailed protocol.

      Helpful?

  4. What is the difference between Ficoll 400 and Ficoll PM 400?

    1 answer
    1. The difference between Ficoll 400 and Ficoll PM 400 is the PM 400 has stricter specifications on the number of microorganisms/g and the endotoxin level is < 0.125 EU/ml

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Can Product F2637, Ficoll, be used as a component of a DNA loading buffer?

    1 answer
    1. This product can be used in a DNA loading buffer with the following recipe to prepare a 6X stock:0.25% bromophenol blue0.25% xylene cyanol FF15% Ficoll

      Helpful?

Reviews

No rating value

Active Filters