Skip to Content
Merck
All Photos(1)

Key Documents

P7458

Sigma-Aldrich

PKH26 Reference Microbeads

Synonym(s):

Red PKH reference beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

storage condition

dry at room temperature

Quality Level

detection method

fluorometric

shipped in

wet ice

storage temp.

2-8°C

General description

PKH26 Reference Microbeads have a fluorescence intensity and light scatter characteristics similar to PKH26-labeled leukocytes. The reference microbeads are impregnated with PKH26 dye, then carefully formulated as a suspension to serve as a fluorescence intensity and count standard.

Application

PKH26 Reference Microbeads has been used:
  • as an internal counting control in flow cytometric acquisition and analysis
  • for viable cell count analysis of fibroblasts
  • for measuring the leukemic cell count by flow cytometry

Linkage

For additional technical details on PKH Fluorescent Cell Linker Dyes including a comprehensive bibliography, please visit here.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam L Burrack et al.
Cell reports, 28(8), 2140-2155 (2019-08-23)
Pancreatic ductal adenocarcinoma (PDA) is a lethal cancer resistant to immunotherapy. We create a PDA mouse model and show that neoantigen expression is required for intratumoral T cell accumulation and response to immune checkpoint blockade. By generating a peptide:MHC tetramer, we
Kah Teong Soh et al.
Cytometry. Part B, Clinical cytometry, 98(2), 161-173 (2019-12-24)
Recent advances in therapeutic interventions have dramatically improved complete response rates in patients with multiple myeloma (MM). The ability to identify residual myeloma cells (e.g., measurable residual disease [MRD]) can provide valuable information pertaining to patient's depth of response to
Mark J Osborn et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 21(6), 1151-1159 (2013-04-03)
Recessive dystrophic epidermolysis bullosa (RDEB) is characterized by a functional deficit of type VII collagen protein due to gene defects in the type VII collagen gene (COL7A1). Gene augmentation therapies are promising, but run the risk of insertional mutagenesis. To
Christian Sordo-Bahamonde et al.
Cancers, 13(9) (2021-05-01)
The inclusion of monoclonal antibodies targeting immune checkpoints such PD-1/PD-L1 or CTLA-4 has revolutionized the landscape of anti-cancer therapy. However, PD-1 and CTLA-4 blockade failed to achieve clinical benefit in CLL, thus attention has been focused on emerging checkpoints in
Jinguo Chen et al.
Toxicology, 249(2-3), 194-203 (2008-07-05)
Low-nicotine and nicotine-free cigarettes are commercially available under the brand-name Quest. Some consumers may believe that these are safer cigarettes, and they may smoke more cigarettes or inhale more smoke to compensate for low nicotine yields. Thus, we have studied

Articles

PKH and CellVue® Fluorescent Cell Linker Kits provide fluorescent labeling of live cells over an extended period of time, with no apparent toxic effects.

PKH and CellVue® Fluorescent Cell Linker Kits provide fluorescent labeling of live cells over an extended period of time, with no apparent toxic effects.

PKH and CellVue® Fluorescent Cell Linker Kits provide fluorescent labeling of live cells over an extended period of time, with no apparent toxic effects.

PKH and CellVue® Fluorescent Cell Linker Kits provide fluorescent labeling of live cells over an extended period of time, with no apparent toxic effects.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service