Skip to Content
Merck
All Photos(2)

Key Documents

31149

Sigma-Aldrich

Deoxyribonucleic acid, low molecular weight from salmon sperm

Synonym(s):

Salmon sperm dsDNA, low molecular weight DNA, DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106310
eCl@ss:
32160414
NACRES:
NA.52

grade

for molecular biology
puriss. p.a.

form

lyophilized powder

impurities

≤5% protein

solubility

50 mM Tris pH 8.0: 10 mg/mL, clear, faintly brown-yellow

absorbance ratio

A260/280 nm ~1.4

storage temp.

2-8°C

Application

Various concentrations of Deoxyribonucleic acid, low molecular weight, from salmon sperm has been used:
  • to study DNA binding to silicon-substituted hydroxyapatite nanocrystals.
  • in simple electrochemical DNA biosensors to detect and evaluate DNA damage caused by UV-C radiation and reactive oxygen species.
  • as a precursor in the synthesis of the bio-dots.

Biochem/physiol Actions

Salmon sperm DNA is often used as a blocking agent invarious research applications. It isknown to improve the specificity and sensitivity of the multi-AP assay byreducing non-specific binding and background signal.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Gao et al.
Journal of immunology (Baltimore, Md. : 1950), 166(11), 6855-6860 (2001-05-22)
LPS is well recognized for its potent capacity to activate mouse macrophages to produce TNF-alpha, an important inflammatory mediator in bacterial infection-related diseases such as septic shock. We demonstrate here that while inducing only low levels of TNF-alpha alone, DNA
Eiji Sunami et al.
Clinical chemistry, 55(3), 559-567 (2009-01-10)
Prostate cancer (PCa) detection using serum-based prostate specific antigen (PSA) is limited by frequent false-positive and -negative results. Genetic aberrations such as allelic imbalance (AI) and epigenetic changes such as promoter hypermethylation have been detected in circulating DNA of cancer
Andrea Hájková et al.
Bioelectrochemistry (Amsterdam, Netherlands), 116, 1-9 (2017-03-18)
A simple electrochemical DNA biosensor based on a glassy carbon electrode (GCE) was prepared by adsorbing double-stranded DNA (dsDNA) onto the GCE surface and subsequently used for the detection of dsDNA damage induced by hydroxyl radicals. Investigation of the mutual
V N Kondratova et al.
Biochemistry. Biokhimiia, 74(11), 1285-1288 (2009-11-18)
A new method of electrophoresis (isotachophoresis in agarose gel rods) in which nucleic acid molecules are not separated but, oppositely, are brought together into one band, was elaborated. Heterogeneous in size DNA and RNA polymers present in a few milliliters
Thiol modification of silicon-substituted hydroxyapatite nanocrystals facilitates fluorescent labelling and visualisation of cellular internalisation
Williams RL et al
Journal of Material Chemistry B: Materials for Biology and Medicine, 35, 4370-4378 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service