Skip to Content
Merck

Skip To

P6136

Brij® O10

Synonym(s):

Brij 97, C18-1E10, Polyoxyethylene (10) oleyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C18H35(OCH2CH2)nOH, n~10
CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

mol wt

709.00 g/mol

density

1 g/mL at 25 °C (lit.)

HLB

12.4

SMILES string

O(CCO)CCCCCCCC\C=C/CCCCCCCC

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

Application

Brij O10 has been used in a study to assess the aqueous surfactant two-phase systems for the continuous countercurrent cloud point extraction.

Legal Information

Brij is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
436240STS0214388866
density

1 g/mL at 25 °C (lit.)

density

-

density

0.901 g/cm3 at 20 °C

density

0.912 g/mL at 25 °C (lit.)

HLB

12.4

HLB

15

HLB

15.5

HLB

4

mol wt

709.00 g/mol

mol wt

average Mn ~1,150

mol wt

average Mn ~1,150

mol wt

average Mn ~357

description

non-ionic

description

non-ionic

description

non-ionic

description

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

>464.0 °F - Equilibrium method

flash_point_c

> 240 °C - Equilibrium method

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Aqueous Surfactant Two-Phase Systems for the Continuous Countercurrent Cloud Point Extraction
Ingram, T., et al.
Chemie Ingenieur Technik, 84, 840-840 (2012)
Abla Benleulmi-Chaachoua et al.
Cellular and molecular life sciences : CMLS, 75(23), 4357-4370 (2018-07-26)
Melatonin, a neuro-hormone released by the pineal gland, has multiple effects in the central nervous system including the regulation of dopamine (DA) levels, but how melatonin accomplishes this task is not clear. Here, we show that melatonin MT1 and MT2
Chiwei Xu et al.
Cell reports, 26(3), 670-688 (2019-01-17)
Hippo signaling and the activity of its transcriptional coactivator, Yorkie (Yki), are conserved and crucial regulators of tissue homeostasis. In the Drosophila midgut, after tissue damage, Yki activity increases to stimulate stem cell proliferation, but how Yki activity is turned



Global Trade Item Number

SKUGTIN
P6136-1KG04061834392972
P6136-500G04061834392989
P6136-100G04061834392965

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Bonjour Faut-il particulièrement protéger ce produit de la lumière ? Y a t'il un tampon ou pH conseillé pour des dilutions stables ?

    1 answer
    1. This product is not particularly sensitive to light but is susceptible to oxidation. The pH limits have not been established. Please see the link below to review the product datasheet: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/263/219/p6136pis.pdf

      Helpful?

Reviews

No rating value

Active Filters