P4019
Brij® S20
Synonym(s):
Polyethylene glycol octadecyl ether, Polyoxyethylene (20) stearyl ether
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C18H37(OCH2CH2)nOH, n~20
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
solid
Quality Level
mp
44-46 °C (lit.)
HLB
15
SMILES string
CCCCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
InChI
1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3
InChI key
PSEGVHKUPXNGGJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Brij® S20 is a surfactant that belongs to the class of polyoxyethylene ethers. It is majorly used in pharmaceutical applications to improve the solubility of the drugs. It can also be grafted on a variety of surfaces to enhance the intracellular uptake of dyes.
Application
- Nanostructured lipid carriers for oral delivery of silymarin: Improving its absorption and in vivo efficacy in type 2 diabetes and metabolic syndrome model.: This study utilizes Brij® S20 to develop nanostructured lipid carriers for oral delivery of silymarin. The findings demonstrate enhanced absorption and efficacy of silymarin in treating type 2 diabetes and metabolic syndrome, highlighting the potential of Brij® S20 in improving drug delivery systems. (Piazzini et al., 2019).
- Water-stable all-biodegradable microparticles in nanofibers by electrospinning of aqueous dispersions for biotechnical plant protection.: The study uses Brij® S20 to create water-stable, biodegradable microparticles in nanofibers through electrospinning. These microparticles are designed for biotechnical plant protection, demonstrating the potential of Brij® S20 in agricultural biotechnology. (Bansal et al., 2012).
Legal Information
Brij is a registered trademark of Croda International PLC
comparable product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nanostructured lipid carriers as promising delivery systems for plant extracts: The case of silymarin
Piazzini V, et al.
Applied Sciences, 8(7), 1163-1163 (2018)
Brij-grafted-chitosan copolymers with function of P-glycoprotein modulation: Synthesis, characterization and in vitro investigations
Xiong W, et al.
Carbohydrate Polymers, 204, 89-96 (2019)
Wei Xiong et al.
International journal of nanomedicine, 13, 3781-3793 (2018-07-11)
Berberine (BBR) is a plant-derived benzylisoquinoline alkaloid and has been demonstrated to be a potential treatment for various chronic diseases. The poor water solubility and P-glycoprotein (Pgp)-mediated drug efflux are the main challenges for its further application in a clinical
L Realini et al.
Journal of clinical microbiology, 35(11), 2791-2794 (1997-11-14)
We report on the influences of polyoxyethylene stearate (POES), PANTA, and pH on primary cultures of Mycobacterium genavense in BACTEC vials. As a model for primary cultures from tissue, seven different strains first isolated from AIDS patients (five from Switzerland
S Rahima Benhabbour et al.
Journal of controlled release : official journal of the Controlled Release Society, 158(1), 63-71 (2011-11-01)
Lipid-based oil-filled nanoparticles (NPs) with a high concentration of surface-chelated nickel (Ni-NPs) were successfully prepared using a Brij 78-NTA-Ni conjugate synthesized with Brij 78 (Polyoxyethylene (20) stearyl ether) and nitrilotriacetic acid (NTA). The facile incorporation of the Brij 78-NTA-Ni conjugate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service