Skip to Content
Merck
All Photos(1)

Documents

P3371

Sigma-Aldrich

Penfluridol

≥97% (HPLC), powder

Synonym(s):

1-[4,4-Bis(4-fluorophenyl)butyl]-4-[4-chloro-3(trifluoromethyl)phenyl]-4-piperidinol, R16341

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H27ClF5NO
CAS Number:
Molecular Weight:
523.97
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white

solubility

DMSO: >20 mg/mL
H2O: insoluble

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

OC1(CCN(CCCC(c2ccc(F)cc2)c3ccc(F)cc3)CC1)c4ccc(Cl)c(c4)C(F)(F)F

InChI

1S/C28H27ClF5NO/c29-26-12-7-21(18-25(26)28(32,33)34)27(36)13-16-35(17-14-27)15-1-2-24(19-3-8-22(30)9-4-19)20-5-10-23(31)11-6-20/h3-12,18,24,36H,1-2,13-17H2

InChI key

MDLAAYDRRZXJIF-UHFFFAOYSA-N

Application

Penfluridol has been used as an antipsychotic agent:
  • to study its antimetastatic effect on triple-negative breast cancer cells
  • to study its effects on the growth of glioblastoma cell lines,
  • to study its effects on the vascular endothelial growth factor (VEGF)-induced angiogenesis in human umbilical vein endothelial cells (HUVECs)

Biochem/physiol Actions

Penfluridol is studied in the treatment of schizophrenia, Tourette syndrome and acute psychosis. It also exhibits anticancer activity.
T-type Ca2+ channel blocker; antipsychotic

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Won et al.
Endocrinology, 126(2), 849-857 (1990-02-01)
We examined the effects of removing extracellular Ca2+ (Ca2+e), depleting intracellular Ca2+ (Ca2+i), inhibiting Ca2(+)-dependent calmodulin, blocking voltage-sensitive Ca2+ channels, and combining Ca2+i depletion with exposure to glucocorticoid on the secretion of ACTH by perifused dispersed rat anterior pituitary cells
J J Enyeart et al.
Molecular pharmacology, 42(2), 364-372 (1992-08-01)
We have used the whole-cell version of the patch-clamp technique to analyze the inhibition of Ca2+ currents by antipsychotic agents in neural crest-derived rat and human thyroid C cell lines. Diphenylbutylpiperidine (DPBP) antipsychotics, including penfluridol and fluspirilene, potently and preferentially
Suyash Srivastava et al.
International journal of molecular sciences, 21(3) (2020-01-26)
: Metastasis is considered a major burden in cancer, being responsible for more than 90% of cancer-related deaths. Tumor angiogenesis is one of the main processes that lead to tumor metastasis. Penfluridol is a classic and commonly used antipsychotic drug
B Spivak et al.
International clinical psychopharmacology, 11(3), 207-209 (1996-09-01)
Neuroleptic malignant syndrome (NMS) is a severe side-effect of neuroleptic treatment. It is usually related to hypodopaminergic activity. A young schizophrenic patient who developed a typical episode of NMS during abrupt withdrawal of long-acting neuroleptic combined with anticholinergic treatment is
M Mungthin et al.
The American journal of tropical medicine and hygiene, 60(3), 469-474 (1999-08-31)
The drug sensitivity characteristics and Plasmodium falciparum pfmdr1 status of five isolates of P. falciparum recently isolated from patients presenting for treatment from the Thailand/Myanmar border have been investigated. The aim of the study was to avoid the criticisms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service