Skip to Content
Merck
All Photos(5)

Documents

138576

Sigma-Aldrich

1-Adamantylamine

97%

Synonym(s):

1-Aminoadamantane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N
CAS Number:
Molecular Weight:
151.25
Beilstein:
2204333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

206-208 °C (lit.)

solubility

1 M HCl: soluble 5%, clear to hazy, colorless to faint yellow or tan

SMILES string

NC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C10H17N/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6,11H2/t7-,8+,9-,10-

InChI key

DKNWSYNQZKUICI-CHIWXEEVSA-N

Gene Information

human ... GRIN2A(2903)

Looking for similar products? Visit Product Comparison Guide

Application

1-Adamantylaminecan be used as a reactant to synthesize:
  • Adamantyl ureas by reacting with isocyanates in the presence of triethylamine.
  • N-(1-adamantyl)-2-chloroacetamide from chloroacetyl chloride in the presence of potassium carbonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jinglun Huang et al.
The Journal of organic chemistry, 72(1), 204-208 (2006-12-30)
The enantioselective Strecker reaction of N-diphenylphosphinoyl ketoimines has been achieved by use of in situ prepared chiral N,N'-dioxide catalyst from l-piperidinamide 3f and m-chloroperoxybenzoic acid (m-CPBA). Excellent yields (up to 99%) and high enantioselectivities (up to 92% ee) were obtained.
Lei Deng et al.
Nature communications, 9(1), 359-359 (2018-01-26)
Current influenza vaccines provide limited protection against circulating influenza A viruses. A universal influenza vaccine will eliminate the intrinsic limitations of the seasonal flu vaccines. Here we report methodology to generate double-layered protein nanoparticles as a universal influenza vaccine. Layered
Terminal Modification with 1-Adamantylamine to Endow Hyperbranched Polyamidoamine with Thermo-/pH-Responsive Properties.
Guo Z, et al.
Macromolecular Rapid Communications, 29(21), 1746-1751 (2008)
Márió Gajdács et al.
Antibiotics (Basel, Switzerland), 8(4) (2019-12-22)
Background: The emergence of multidrug-resistant organisms (MDROs) is a global public health issue, severely hindering clinicians in administering appropriate antimicrobial therapy. Drug repurposing is a drug development strategy, during which new pharmacological applications are identified for already approved drugs. From
Dinesh Addla et al.
Bioorganic & medicinal chemistry letters, 24(8), 1974-1979 (2014-04-01)
A series of novel 1,2,3-triazole-adamantylacetamide hybrids 5a-u, designed by combining bioactive fragments from antitubercular I-A09 and substituted adamantyl urea, were synthesized using copper catalyzed click chemistry. N-(1-Adamantyl)-2-azido acetamide 3 prepared from 1-adamantylamine was reacted with a series of alkyl/aryl acetylenes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service