Skip to Content
Merck
All Photos(3)

Key Documents

P0501

Sigma-Aldrich

Phenolphthalein β-D-glucuronide

β-glucuronidase substrate

Synonym(s):

Phenolphthalein β-D-glucosiduronic acid, Phenolphthalein glucuronic acid, Phenolphthalein mono-β-D-glucosiduronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O10
CAS Number:
Molecular Weight:
494.45
Beilstein:
68526
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (TLC)

form

powder

solubility

H2O: soluble 100 mg/mL

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5

InChI

1S/C26H22O10/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32/h1-12,19-22,25,27-30H,(H,31,32)/t19-,20-,21+,22-,25+,26?/m0/s1

InChI key

FXJYOZKDDSONLX-XADSOVDISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

β-glucuronidase activity was determined by the amount of phenolphthalein released from 0.01 M henolphthalein β-D-glucuronide.
Phenolphthalein β-D-glucuronide has been used as a substrate for the detection of β-D-glucuronidase activity in fecal bacteria.

Biochem/physiol Actions

Phenolphthalein β-D-glucuronide is a chromogenic substrate for β-glucuronidase enzyme and the resultant colored product of the reaction is measured at 552 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A microtitre plate assay for measuring glycosidase activity
Ball Al, et al.
Journal of Enzyme Inhibition and Medicinal Chemistry, 23(1), 131-135 (2008)
Effects of two pomace insoluble fibres on the activities of faecal bacterial enzymes and intestinal health
Chau CF and Chen CH
European Food Research and Technology, 222(5-6), 681-685 (2006)
Hisashi Satoh et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 83(6), 1399-1406 (2021-03-27)
Monitoring of Escherichia coli concentrations in river water (RW) is essential to identify fecal pollution of the river. The objective of this study was to assess the suitability of a novel, simple and high throughput method developed in our laboratory
Improvement in intestinal function of hamsters as influenced by consumption of polysaccharide-rich sage weed extracts
Huang YL and Chau CF
Food Chemistry, 133(4), 1618-1623 (2012)
Intestinal health benefits of the water-soluble carbohydrate concentrate of wild grape (Vitis thunbergii) in hamsters
Huang YL, et al.
Journal of Agricultural and Food Chemistry, 60(19), 4854-4858 (2012)

Articles

Drug conjugate analysis and the enzymatic hydrolysis of glucuronides

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

See All

Protocols

Objective: To standardize a procedure for the enzymatic assay of β-Glucuronidase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service