Skip to Content
Merck
All Photos(1)

Documents

N2783

Sigma-Aldrich

Nitric Oxide Synthase, Inducible from mouse

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

Inducible Nitric Oxide Synthase, NOS II, iNOS, macNOS

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥4.0 units/mg protein

mol wt

130 kDa (homodimer)
130 kDa (subunit, homodimer)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Nos2(18126)

Looking for similar products? Visit Product Comparison Guide

General description

Inducible nitric oxide synthase (iNOS), also known as inflammatory nitric oxide synthase, is a calcium independent isoenzyme, involved in synthesis of nitric oxide (NO). It is a soluble enzyme encoded by the gene mapped to mouse chromosome 11. iNOS is active in dimeric form and its activity is induced by cytokines and various other stimuli. iNOS is expressed in various inflammatory conditions.

Application

Nitric Oxide Synthase, Inducible from mouse has been used in immunohistochemical studies. It is also used to evaluate the therapeutic efficacy of inducible nitric oxide synthase (NOS) on reperfusion-induced microcirculatory alterations and hemodynamic adverse effects in the microvasculature of skeletal muscle.

Biochem/physiol Actions

Tumor-derived inducible nitric oxide synthase (iNOS) plays a vital role in stimulating tumor growth and vessel maturation. Therefore, it is considered to be a potential therapeutic target for anti-vascular cancer therapies. Unchecked activity of iNOS leads to overproduction of nitric oxide (NO), which is toxic for living cells. iNOS activity can be controlled at both transcription and translational level by regulating protein stability, dimerization, phosphorylation, cofactor binding and availability of oxygen and L-arginine as substrates. iNOS plays a vital role in excisional wound repair and exhibits gene therapy strategy to advance wound healing process in iNOS-deficient conditions such as diabetes and steroid treatment.
NOS is responsible for the biosynthesis of nitric oxide from L-arginine. iNOS is not calcium/calmodulin dependent and has a Km = 16 μM for L-arginine.

Unit Definition

One unit will produce 1.0 μmol of nitric oxide per minute at 37 °C in 50 mM HEPES, pH 7.4, containing 1 mM arginine, 1 mM magnesium acetate, 0.15 mM NADPH, 4.5 μM oxyhemoglobin, 18 μM tetrahydrobiopterin and 180 μM DTT.

Physical form

Solution in 50 mM HEPES, pH 7.4, with 10% glycerol, 8 μM tetrahydrobiopterin

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigating the role of tumour cell derived iNOS on tumour growth and vasculature in vivo using a tetracycline regulated expression system.
Papaevangelou E
International Journal of Cancer. Journal International Du Cancer, 138(11), 2678-2687 (2016)
Mapping of the gene for inducible nitric oxide (NO) synthase of mouse macrophages to chromosome 11, close to Evi-2, nu, and Idd-4.
Jenkins NA
Genomics, 19(2), 402-404 (1994)
Enhancing Nitric Oxide Bioavailability via Exogen Nitric Oxide Synthase and L-Arginine Attenuates Ischemia-Reperfusion-Induced Microcirculatory Alterations.
Engel H
Annals of Plastic Surgery (2014)
iNOS-mediated nitric oxide production and its regulation.
Aktan F
Life Sciences, 75(6), 639-653 (2004)
Reversal of impaired wound repair in iNOS-deficient mice by topical adenoviral-mediated iNOS gene transfer.
Yamasaki K
The Journal of Clinical Investigation, 101(5), 967-971 (1998)

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service