Skip to Content
Merck
All Photos(2)

Key Documents

N2640

Sigma-Aldrich

4-Nitrophenyl phosphate disodium salt hexahydrate

chromogenic, tablet

Synonym(s):

p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
Beilstein:
5324661
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

4-Nitrophenyl phosphate disodium salt hexahydrate, tablet

form

tablet

solubility

water: 1 tablet/3 mL, clear, colorless

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chromogenic substrate for the determination of acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm.

Application

4-Nitrophenyl phosphate disodium salt hexahydrate has been used in enzyme-linked immunosorbent assay (ELISA) as a substrate for streptavidin-Alkaline Phosphatase. It has also been used as a substrate in control assay for comparing 4-aminophenyl phosphate/diethanolamine (APP/DEA) assay for alkaline phosphatase.

Biochem/physiol Actions

4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) is a non-proteinaceous and non-specific substrate for determining protein tyrosine phosphatase activity. Hydrolysis of pNPP yields p-nitrophenol. pNPP is a common chromogenic substrate for alkaline phosphatase.

Quantity

Contains 15 mg substrate per tablet

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin A differentially regulates cytokine expression in respiratory epithelial and macrophage cell lines
Penkert RR, et al.
Cytokine, 91, 1-5 (2017)
Christopher J Sample et al.
Peptides, 48, 96-105 (2013-07-31)
Broad-spectrum antiviral drugs are urgently needed to treat individuals infected with new and re-emerging viruses, or with viruses that have developed resistance to antiviral therapies. Mammalian natural host defense peptides (mNHP) are short, usually cationic, peptides that have direct antimicrobial
Alkaline Phosphatase Assay Based on the Chromogenic Interaction of Diethanolamine with 4-Aminophenol
Sun J, et al.
Analytical Chemistry, 90(10), 6339-6345 (2018)
Protein tyrosine phosphatase assays
Lorenz U
Current Protocols in Immunology, 93(1), 11-17 (2011)
Christina Papista et al.
Kidney international, 88(2), 276-285 (2015-03-26)
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service