Skip to Content
Merck

N3129

4-Nitrophenyl phosphate bis(cyclohexylammonium) salt

phosphatase substrate, chromogenic, ≥97% anhydrous basis, powder

Synonym(s):

p-Nitrophenyl phosphate bis(cyclohexylammonium) salt, PNPP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
O2NC6H4OP(O)(O-)2 · 2C6H11NH3+
CAS Number:
Molecular Weight:
417.44
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
257-949-0
MDL number:
Beilstein/REAXYS Number:
6553276
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

4-Nitrophenyl phosphate bis(cyclohexylammonium) salt, phosphatase substrate

Quality Segment

assay

≥97% anhydrous basis

form

powder

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.OP(O)(=O)Oc3ccc(cc3)[N+]([O-])=O

InChI

1S/C6H6NO6P.2C6H13N/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;2*7-6-4-2-1-3-5-6/h1-4H,(H2,10,11,12);2*6H,1-5,7H2

InChI key

KXNNUYCXODNFBD-UHFFFAOYSA-N

Application

4-Nitrophenyl phosphate bis(cyclohexylammonium) salt has been used as a component in the colorimetric reagent for abscisic acid (ABA) assay.

Biochem/physiol Actions

p-Nitrophenyl phosphate (pNPP) is the preferred substrate for use with alkaline phosphatase in enzyme-linked immunosorbent assay (ELISA) procedures. Due to its high sensitivity, pNPP is the considered substrate in alkaline phosphatase systems. ELISA applications based on pNPP can be read in timed assays or delayed readings by stopping with alkaline solutions. This substrate yields a yellow-colored soluble end product that can be measured spectrophotometrically at 405 nm.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library