MSST0005
SILu™Prot VEGFA Vascular endothelial growth factor A human
recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C- and 15N-labeled
Synonym(s):
SILu™Prot Vascular endothelial growth factor A
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
23201100
NACRES:
NA.12
Recommended Products
biological source
human
Quality Level
recombinant
expressed in HEK 293 cells
Assay
≥95% (SDS-PAGE)
form
lyophilized powder
technique(s)
mass spectrometry (MS): suitable
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... VEGFA(7422)
General description
SILu™ Prot VEGF165 is a recombinant, stable isotope-labeled human VEGF165 which incorporates [13C6,15N4]−Arginine and [13C6, 15N2]−Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of VEGF165 by mass-spectrometry.
Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)
Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)
Biochem/physiol Actions
VEGF165 belongs to the PDGF/VEGF growth factor family characterized by the presence of eight conserved cysteine residues and a cystine knot structure1. VEGF is secreted by the majority of tumor cells and initiates angiogenesis by activating endothelial cells of existing blood vessels and promoting their migration2.
VEGF has also been implicated in correlation with poor prognosis in breast cancer2. In addition, VEGF is released in rheumatoid arthritis in response to TNF-α, increasing endothelial permeability and stimulating angiogenesis (formation of capillaries)3,4.
VEGF has also been implicated in correlation with poor prognosis in breast cancer2. In addition, VEGF is released in rheumatoid arthritis in response to TNF-α, increasing endothelial permeability and stimulating angiogenesis (formation of capillaries)3,4.
Sequence
APMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGC
Physical form
Supplied as a lyophilized powder containing phosphate buffered saline
Legal Information
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice),1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service