Skip to Content
Merck

Skip To

M8937

Greiner CELLSTAR® multiwell culture plates

48 wells (TC treated with lid)

Synonym(s):

culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates, Greiner 677180

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
clear wells, flat clear wells, polystyrene
Size:
48 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear wells, flat clear wells, polystyrene

sterility

sterile

feature

lid

packaging

pack of 1 ea (single packed)

manufacturer/tradename

Greiner 677180

size

48 wells

well volume

1.7 mL

well working volume

0.5- 1.0 mL

color

clear

binding type

Tissue Culture (TC)-treated surface

Features and Benefits

  • CELLSTAR® 48 Well Cell Culture Multiwell Plates enables improved cell adhesion through physical surface treatment.
  • Features a lid that facilitates gas exchange with the lowest possible evaporation and significantly reduces the risk of cross-contamination.
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic.
  • Suitable for all common instruments and compatible with automated systems.
  • Alphanumeric well coding for easy well/sample identification.

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M9437M8687M8562
description

48 wells (TC treated with lid)

description

48 wells (suspension culture with lid)

description

12 wells (TC treated with lid), flat bottom, pack of 100 ea sterile, individually wrapped

description

6 wells (TC treated with lid)

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

manufacturer/tradename

Greiner 677180

manufacturer/tradename

Greiner 677102

manufacturer/tradename

Greiner 665180

manufacturer/tradename

Greiner 657160

feature

lid

feature

lid

feature

lid

feature

lid

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

material

clear wells, polystyrene , flat clear wells

material

flat clear wells, polystyrene

material

flat bottom , flat bottom wells, polystyrene

material

flat clear wells, polystyrene

size

48 wells

size

48 wells

size

12 wells

size

6 wells


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library