Skip to Content
Merck
All Photos(1)

Key Documents

M6074

Sigma-Aldrich

Minimum Essential Medium Eagle

StableCell, Alpha modification, with stable glutamine and sodium bicarbonate, with nucleosides, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

StableCell MEM, Alpha Modification, With stable glutamine and sodium bicarbonate, with ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

Hanks’ salts (2% CO2): no
Earle’s salts (5% CO2): yes
phenol red: yes
glucose: yes
HEPES: no
sodium pyruvate: no
stable glutamine: yes
NaHCO3: yes
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This MEM differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Giunta et al.
The Journal of cell biology, 190(2), 197-207 (2010-07-28)
The signaling cascade initiated in response to DNA double-strand breaks (DSBs) has been extensively investigated in interphase cells. Here, we show that mitotic cells treated with DSB-inducing agents activate a "primary" DNA damage response (DDR) comprised of early signaling events
Pasqualino de Antonellis et al.
PloS one, 6(9), e24584-e24584 (2011-09-21)
Through negative regulation of gene expression, microRNAs (miRNAs) can function as oncosuppressors in cancers, and can themselves show altered expression in various tumor types. Here, we have investigated medulloblastoma tumors (MBs), which arise from an early impairment of developmental processes
Carlos Mendoza-Palomares et al.
Eukaryotic cell, 7(4), 684-697 (2008-02-19)
Cysteine proteases have been shown to be essential virulence factors and drug targets in trypanosomatids and an attractive antidisease vaccine candidate for Trypanosoma congolense. Here, we describe an important amplification of genes encoding cathepsin B-like proteases unique to T. congolense.
Barbara Zonta et al.
Neuron, 69(5), 945-956 (2011-03-09)
The axon initial segment (AIS) is critical for the initiation and propagation of action potentials. Assembly of the AIS requires interactions between scaffolding molecules and voltage-gated sodium channels, but the molecular mechanisms that stabilize the AIS are poorly understood. The
Hisashi Miura et al.
Nucleic acids research, 37(3), 778-792 (2008-12-17)
Mammalian tissue- and/or time-specific transcription is primarily regulated in a combinatorial fashion through interactions between a specific set of transcriptional regulatory factors (TRFs) and their cognate cis-regulatory elements located in the regulatory regions. In exploring the DNA regions and TRFs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service