Skip to Content
Merck

Skip To

S2GPU11RE

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.22 μm, polyethersulfone membrane, filter capacity 1000 mL, pack of 12 ea

Synonym(s):

SCGPU11RE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24
eCl@ss:
32031623
Pore size:
0.22 μm pore size
Filter capacity:
1000 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Sterility:
sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

sterility

sterile

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

319 mm

diam.

13.5 cm

filter capacity

1000 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel, outlet flask (Receiver)

shipped in

ambient

storage temp.

no temp limit

General description

This item is a direct replacement for SCGPU11RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.22 μm pore size polyethersulfone (PES) membrane filter unit combines a 1000 mL bottle-top with a 1000 mL receiver flask and cap for processing and storage.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.
  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2GPU10RES2GPU01RES2GPU02RE
description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 1000 mL, pack of 12 ea

description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, pack of 12 ea

description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 150 mL, pack of 12 ea

description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 250 mL, pack of 12 ea

sterility

sterile

sterility

sterile

sterility

-

sterility

sterile

filter capacity

1000 mL

filter capacity

500 mL

filter capacity

150 mL

filter capacity

250 mL

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

material

polyethersulfone membrane, polystyrene funnel(s), polystyrene bottle, polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 17 Questions  
  1. Can the bottle be autoclaved and reused after storage? I understand that the filter is temperature-sensitive, but what about the bottle itself?

    1 answer
    1. For the S2GPU11RE and all Stericup/Steritop products, these are designed to be one time use only.  We have not studied the effects of autoclaving or reusing this on the plastic.

      Helpful?

  2. ​Is there an integrity test available for the S2GPU11RE?

    1 answer
    1. No, an integrity test is not available for the S2GPU11RE.

      Helpful?

  3. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

  4. What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
      *Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.

      Helpful?

  5. Can Stericup® and Steritop® filters be reused?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single use, disposable processing.

      Helpful?

  6. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  7. Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?

    1 answer
    1. No, all Stericup® and Steritop® devices are Research Use Only.

      Helpful?

  8. What is the expiration date of Stericup® and Steritop® filters?

    1 answer
    1. Typically 3 years from sterilization.

      Helpful?

  9. Can Stericup® bottles be placed inside freezers for long-term storage of media?

    1 answer
    1. You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.

      Helpful?

  10. What leachables are detected during filtration processes using Stericup® filters?

    1 answer
    1. We have not performed extractables/leachables studies for our Stericup® filters.

      Helpful?

1–10 of 17 Questions  

Reviews

Active Filters

  1. 1 Ratings-only review