Skip to Content
Merck
All Photos(4)

Key Documents

M3824

Sigma-Aldrich

α-Mangostin

≥98% (HPLC)

Synonym(s):

α-MG, alpha-Mangostin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O6
CAS Number:
Molecular Weight:
410.46
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

solubility

methanol: 1 mg/mL, clear, very light yellow

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

COc1c(O)cc2Oc3cc(O)c(C\C=C(\C)C)c(O)c3C(=O)c2c1C\C=C(/C)C

InChI

1S/C24H26O6/c1-12(2)6-8-14-16(25)10-19-21(22(14)27)23(28)20-15(9-7-13(3)4)24(29-5)17(26)11-18(20)30-19/h6-7,10-11,25-27H,8-9H2,1-5H3

InChI key

GNRIZKKCNOBBMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Mangostin (α-MG) is the most abundant phytochemical derived from the pericarp of Garcinia mangostana L.. It is a tetraoxygenated diprenylated xanthone. The chemical structure of α-MG molecule is 1,3,6-trihydroxy-7-methoxy-2,8-bis(3-methyl-2-butenyl)- 9H-xanthen-9-one.

Application

α-Mangostin has been used:
  • to study its protective effect against oxidative stress mediated-retinal cell death
  • to study its role as an antigenotoxic agent in DNA protection against oxidative damage
  • to understand its anti-bacterial activity against Staphylococcus epidermidis RP62A

Biochem/physiol Actions

α-Mangostin displays various pharmacological properties both in in vitro and in vivo studies. These include antimicrobial, anticarcinogenic, antidiabetic, neuroprotective, hepatoprotective and anti-inflammatory properties.
Xanthone derivative isolated from mangosteen (Garcinia mangostana). Antioxidant and anti-inflammatory.
Xanthone derivative isolated from mangosteen (Garcinia mangostana). Antioxidant and anti-inflammatory. α-mangostin has been shown to induce apoptosis via the mitochondrial pathway, reduce cell proliferation and inhibit tumorigenesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaofang Quan et al.
PloS one, 7(3), e33376-e33376 (2012-03-20)
α-Mangostin, isolated from the hulls of Garcinia mangostana L., was found to have in vitro cytotoxicity against 3T3-L1 cells as well as inhibiting fatty acid synthase (FAS, EC 2.3.1.85). Our studies showed that the cytotoxicity of α-mangostin with IC(50) value
Ali Ghasemzadeh et al.
Molecules (Basel, Switzerland), 23(8) (2018-07-26)
Since α-mangostin in mangosteen fruits was reported to be the main compound able to provide natural antioxidants, the microwave-assisted extraction process to obtain high-quality α-mangostin from mangosteen pericarp (Garcinia mangostana L.) was optimized using a central composite design and response
Fabiola Gutierrez-Orozco et al.
Journal of agricultural and food chemistry, 61(16), 3891-3900 (2013-04-13)
Information about the anti-inflammatory activity and metabolism of α-mangostin (α-MG), the most abundant xanthone in mangosteen fruit, in human cells is limited. On the basis of available literature, we hypothesized that α-MG will inhibit the secretion of pro-inflammatory mediators by
DNA Protection against Oxidative Damage Using the Hydroalcoholic Extract of Garcinia mangostana and Alpha-Mangostin.
Carvalho-Silva R
Evidence-Based Complementary and Alternative Medicine : ECAM, 2016, 3430405-3430405 (2016)
Jeremy J Johnson et al.
Carcinogenesis, 33(2), 413-419 (2011-12-14)
There is a need to characterize promising dietary agents for chemoprevention and therapy of prostate cancer (PCa). We examined the anticancer effect of α-mangostin, derived from the mangosteen fruit, in human PCa cells and its role in targeting cell cycle-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service