Skip to Content
Merck
All Photos(2)

Key Documents

M8777

Sigma-Aldrich

Morphine sulfate salt pentahydrate

Synonym(s):

Morphine hemi[sulfate pentahydrate]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H40N2O10S · 5H2O
CAS Number:
Molecular Weight:
758.83
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

solubility

ethanol: 1.8 mg/mL
H2O: 64 mg/mL

originator

Baxter

SMILES string

O.O.O.O.O.OS(O)(=O)=O.CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35.CN6CC[C@]78[C@H]9Oc%10c(O)ccc(C[C@@H]6[C@@H]7C=C[C@@H]9O)c8%10

InChI

1S/2C17H19NO3.H2O4S.5H2O/c2*1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;1-5(2,3)4;;;;;/h2*2-5,10-11,13,16,19-20H,6-8H2,1H3;(H2,1,2,3,4);5*1H2/t2*10-,11+,13-,16-,17-;;;;;;/m00....../s1

InChI key

GRVOTVYEFDAHCL-RTSZDRIGSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Narcotic analgesic; prototypic μ opioid receptor agonist; also agonist at κ opioid receptors.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Katsumata et al.
European journal of pharmacology, 291(3), 367-373 (1995-11-30)
We investigated the binding characteristics of dihydroetorphine, 7,8-dihydro-7 alpha-[1-(R)-hydroxy-1-methylbutyl]-6, 14-endoethanotetrahydro-oripavine, and its effect on the inhibitory system of cyclic AMP production using cloned mu-, delta- and kappa-opioid receptors expressed on Chinese hamster ovary cells. The Ki values of dihydroetorphine for
L Stanasila et al.
Journal of neurochemistry, 75(3), 1190-1199 (2000-08-11)
Two constructs encoding the human micro-opioid receptor (hMOR) fused at its C terminus to either one of two Galpha subunits, Galpha(o1) (hMOR-Galpha(o1)) and Galpha(i2) (hMOR-Galpha(i2)), were expressed in Escherichia coli at levels suitable for pharmacological studies (0.4-0.5 pmol/mg). Receptors fused
Stuti Agarwal et al.
American journal of physiology. Lung cellular and molecular physiology, 318(5), L1097-L1108 (2020-04-03)
We previously demonstrated that the combined exposure of human pulmonary microvascular endothelial cells (HPMECs) to morphine and viral protein(s) results in the oxidative stress-mediated induction of autophagy, leading to shift in the cells from early apoptotic to apoptosis-resistant proliferative status
G Martin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(20), 9081-9089 (1999-10-12)
In a study of a possible substrate underlying morphine addiction, we examined NMDA receptor-mediated synaptic transmission of core nucleus accumbens neurons after chronic morphine treatment, using intracellular recording in a slice preparation of rat. We evoked pharmacologically isolated NMDA EPSCs
Serotonin, morphine, and neuropathic pain: not a simple story.
Kelly Byrne et al.
Anesthesiology, 121(2), 217-218 (2014-06-03)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service