Skip to Content
Merck
All Photos(1)

Key Documents

L5520

Sigma-Aldrich

L-15 Medium (Leibovitz)

Without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Leibovitz L15

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: 0.011 g/L
sodium pyruvate: 0.55 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

L-15 Medium (Leibovitz) was initially prepared to be used in carbon dioxide (CO2) free systems, which involves sodium bicarbonate. The complement of free base amino acids, salts, and galactose buffers it. This aids the maintenance of physiological pH control.

Application

L-15 Medium (Leibovitz) has been used:  
  • in protein incorporation for incubation of high blastula embryos and to avoid the abnormal development of embryos
  • as a supplement in sterile cell media for the dissection and isolation of kidney cells
  • as a component of culture buffer for the tissue culture of Newt lung cells

Reconstitution

Supplement with 0.3 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daichi Kayo et al.
Scientific reports, 9(1), 8868-8868 (2019-06-22)
In vertebrates, sex steroids play crucial roles in multiple systems related to reproduction. In females, estrogens and their receptor estrogen receptor (ER or Esr) play indispensable roles in the negative sex steroid feedback regulation of pituitary gonadotropin secretion, which prevents
Olga Krylyshkina et al.
The Journal of cell biology, 156(2), 349-359 (2002-01-25)
Recent studies have shown that the targeting of substrate adhesions by microtubules promotes adhesion site disassembly (Kaverina, I., O. Krylyshkina, and J.V. Small. 1999. J. Cell Biol. 146:1033-1043). It was accordingly suggested that microtubules serve to convey a signal to
Francisco J Monje et al.
Annals of medicine, 45(4), 301-307 (2013-05-02)
Studies of synaptic plasticity using the marine mollusk Aplysia californica as model system have been successfully used to identify proteins involved in learning and memory. The importance of molecular elements regulated by the learning- related neurotransmitter serotonin in Aplysia can
Applications of cell lines as bioreactors and in vitro models
Singh J and Goswami A
International journal of applied & basic medical research null
B Houchmandzadeh et al.
The Journal of cell biology, 139(1), 1-12 (1997-10-06)
The structure of mitotic chromosomes in cultured newt lung cells was investigated by a quantitative study of their deformability, using micropipettes. Metaphase chromosomes are highly extensible objects that return to their native shape after being stretched up to 10 times

Articles

L-15 Media Formulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service