Skip to Content
Merck

Skip To

I5904

Iberiotoxin, recombinant from Mesobuthus tamulus

≥98% (HPLC), potassium channel inhibitor

Synonym(s):

IbTX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C179H276N50O56S7
Molecular Weight:
4248.86
UNSPSC Code:
12352200
NACRES:
NA.77
PubChem Substance ID:
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Iberiotoxin, recombinant from Mesobuthus tamulus, ≥98% (HPLC), recombinant, expressed in E. coli

recombinant

expressed in E. coli

Quality Segment

assay

≥98% (HPLC)

storage temp.

−20°C

Application

Iberiotoxin, recombinant from Mesobuthus tamulus has been used as an inhibitor of large-conductance calcium-activated K+ channels to study its effect on the NLRP3 (nucleotide-binding oligomerization domain-like receptor containing pyrin domain 3) inflammasome activation in bone marrow-derived macrophages (BMDMs). It has also been used to block Ca2+- activated K+ channels to determine interleukin(IL)-1β in BMDCs following inflammasome activation.

Biochem/physiol Actions

Exhibits the same activity as the natural form of the toxin
The single chain peptide iberiotoxin (IbTX) is a selective and reversible inhibitor of high-conductance calcium-activated potassium channels (PK,Ca). It occurs naturally in the venom of the scorpion Buthus tamulus. IbTX also modulates the binding of charybdotoxin (CbTX) to smooth muscle sarcolemmal membranes in a non-competitive manner. IbTX is similar in size to CbTX and shares considerable sequence homology with CbTX. However, IbTX differs in its overall charge in having one fewer basic and four more acidic residues than CbTX.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Soluble in aqueous buffers. Stock solutions of 1 μM can be aliquoted and stored at -20 °C for up to 3 months.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
401002M2547410957
description

≥98% (HPLC), recombinant, expressed in E. coli

description

-

description

≥98%

description

A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC50 = 2-50 µM).

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98%

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

recombinant

expressed in E. coli

recombinant

-

recombinant

-

recombinant

-


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters