Skip to Content
Merck

401002

Iberiotoxin, Buthus tamulus, Recombinant, E. coli

Synonym(s):

Iberiotoxin, Buthus tamulus, Recombinant, E. coli, rlbTx, Pyr-FTDVDCSVSKECWSVCKDLFGVDRGKCMGKKCRCYQ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
lyophilized solid (Salt-free)
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

form

lyophilized solid (Salt-free)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

SMILES string

S1SCC(C(=O)NC(C(=O)NC(C1)C(=O)NC(Cc9ccc(cc9)O)C(=O)NC(CCC(=O)N)C(=O)O)CCCN\C(=N\[H])\N)NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C2NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(CSSC2)NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C6NC(=O

InChI

1S/C179H274N50O55S7/c1-87(2)65-111-155(261)208-112(66-93-33-15-13-16-34-93)146(252)195-77-133(239)225-139(88(3)4)172(278)213-117(71-136(244)245)158(264)199-100(44-31-62-190-178(186)187)144(250)193-75-131(237)198-102(40-22-27-58-181)148(254)219-124(167(273)205-109(56-64-285-12)145(251)194-76-132(238)197-101(39-21-26-57-180)147(253)200-105(43-25-30-61-184)151(257)220-123-81-286-288-83-125(221-152(258)106(203-166(123)272)45-32-63-191-179(188)189)169(275)210-113(68-95-46-48-97(234)49-47-95)156(262)206-110(177(283)284)50-53-129(185)235)82-287-291-86-128(168(274)202-104(42-24-29-60-183)150(256)212-116(70-135(242)243)159(265)207-111)224-175(281)142(91(9)10)228-164(270)121(79-231)216-157(263)115(69-96-74-192-99-38-20-19-37-98(96)99)211-170(276)126-84-289-290-85-127(171(277)217-122(80-232)165(271)227-141(90(7)8)174(280)218-120(78-230)163(269)201-103(41-23-28-59-182)149(255)204-108(154(260)222-126)52-55-134(240)241)223-160(266)118(72-137(246)247)214-173(279)140(89(5)6)226-162(268)119(73-138(248)249)215-176(282)143(92(11)233)229-161(267)114(67-94-35-17-14-18-36-94)209-153(259)107-51-54-130(236)196-107/h13-20,33-38,46-49,74,87-92,100-128,139-143,192,230-234H,21-32,39-45,50-73,75-86,180-184H2,1-12H3,(H2,185,235)(H,193,250)(H,194,251)(H,195,252)(H,196,236)(H,197,238)(H,198,237)(H,199,264)(H,200,253)(H,201,269)(H,202,274)(H,203,272)(H,204,255)(H,205,273)(H,206,262)(H,207,265)(H,208,261)(H,209,259)(H,210,275)(H,211,276)(H,212,256)(H,213,278)(H,214,279)(H,215,282)(H,216,263)(H,217,277)(H,218,280)(H,219,254)(H,220,257)(H,221,258)(H,222,260)(H,223,266)(H,224,281)(H,225,239)(H,226,268)(H,227,271)(H,228,270)(H,229,267)(H,240,241)(H,242,243)(H,244,245)(H,246,247)(H,248,249)(H,283,284)(H4,186,187,190)(H4,188,189,191)

InChI key

VDNVVLOBNHIMQA-UHFFFAOYSA-N

General description

Recombinant Buthus tamulus iberiotoxin expressed in E. coli. A single polypeptide chain of 37 amino acids that displays about 68% sequence homology with charybdotoxin (ChTx). Acts as a selective, reversible blocker of high conductance, Ca2+-activated K+ channels (PK,Ca). rIbTx does not block other types of voltage-dependent ion channels, especially other types of K+ channels that are sensitive to inhibition by ChTx. Shown to block PK,Ca in excised membrane patches from bovine aortic smooth muscle, acting exclusively at the outer face of the channel (IC50 = 250 pM). Causes a decrease in probability of channel opening and the channel open mean time. rIbTx appears to modulate ChTx binding by an allosteric mechanism.

Biochem/physiol Actions

Cell permeable: no
Primary Target
High conductance Ca2+-activated K+ channels
Product does not compete with ATP.
Reversible: yes
Target IC50: 250 pM against high conductance Ca2+-activated K+ channels
potently inhibits Ca2+-activated K+ channels expressed in Xenopus oocytes

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 3 months at -20°C or up to 2 weeks at 4°C.
Reconstitute in Tris-buffered saline (100 mM NaCl, 10 mM Tris-HCl, pH 7.5) containing 0.1% BSA.

Other Notes

Pyr-Phe-Thr-Asp-Val-Asp-Cys⁷-Ser-Val-Ser-Lys-Glu-Cys¹³-Trp-Ser-Val-Cys¹⁷-Lys-Asp-Leu-Phe-Gly-Val-Asp-Arg-Gly-Lys-Cys²⁸-Met-Gly-Lys-Lys-Cys³³-Arg-Cys³⁵-Tyr-Gln-OH (disulfide bonds: 7 →28; 13 → 35; 17 → 33)
Vanelli, G., et al. 1994. J. Appl. Physiol.76, 1098.
Garcia, M.L., et al. 1991. J. Bioeng. Biomembr.23, 615.
Galvez, A., et al. 1990. J. Biol. Chem. 265, 11083.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library