Skip to Content
Merck
All Photos(1)

Key Documents

I5154

Sigma-Aldrich

IgG1, Kappa from human myeloma plasma

purified immunoglobulin, >95% (SDS-PAGE)

Synonym(s):

Human IgG1-κ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

human

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

Assay

>95% (SDS-PAGE)

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.[1] IgG1 is the most abundant of the IgG subclass
Human myeloma IgG1, κ is purified from human plasma by fractionation, ion-exchange, and affinity chromatography procedures.

Application

The purified IgG1, κ may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays. Human myeloma IgG1, κ was injected into mice (500 μg) to assess the T cell and B cell responses[2] and also used for flow cytometry in CHO cells.[3]

Physical form

Solution in 0.02 M Tris buffered saline, pH 8.0.

Analysis Note

Identity is determined by immunoelectrophoresis and indirect ELISA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Y Choi et al.
Cell stem cell, 22(6), 951-959 (2018-06-03)
Cirmtuzumab is a humanized monoclonal antibody (mAb) that targets ROR1, an oncoembryonic orphan receptor for Wnt5a found on cancer stem cells (CSCs). Aberrant expression of ROR1 is seen in many malignancies and has been linked to Rho-GTPase activation and cancer
Johnny W Peterson et al.
Infection and immunity, 74(2), 1016-1024 (2006-01-24)
Prevention of inhalation anthrax requires early and extended antibiotic therapy, and therefore, alternative treatment strategies are needed. We investigated whether a human monoclonal antibody (AVP-21D9) to protective antigen (PA) would protect mice, guinea pigs, and rabbits against anthrax. Control animals
Bas Cornelis Jansen et al.
PloS one, 13(7), e0200280-e0200280 (2018-07-07)
High-performance liquid chromatography (HPLC) is widely used for absolute quantitation. The advent of new columns and HPLC technology has enabled higher sample throughput, and hence, larger scale studies that perform quantitation on different sample types (e.g. healthy controls vs. patients
Yann Gallais et al.
Immunology and cell biology, 95(3), 306-315 (2016-10-08)
Patients treated with therapeutic biological products (BP) frequently develop anti-drug antibodies (ADA) with potential neutralizing capacities leading to loss of clinical response or serious side effects. BP aggregates have been suggested to promote immunogenicity, thus enhancing ADA production. Dendritic cells
Taehong Kwon et al.
Analytical chemistry, 92(7), 5267-5275 (2020-03-14)
We demonstrate a new micro/nanofluidic system for continuous and automatic monitoring of protein product size and quantity directly from the culture supernatant during a high-cell-concentration CHO cell perfusion culture. A microfluidic device enables clog-free cell retention for a bench-scale (350

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service