Skip to Content
Merck

Skip To

I5029

IgG1, Lambda from human myeloma plasma

purified immunoglobulin, >95% (SDS-PAGE)

Synonym(s):

Human IgG1-λ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Species reactivity:
-
Application:
—
Citations:
18
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

assay

>95% (SDS-PAGE)

shipped in

dry ice

storage temp.

−20°C

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. IgG1 is the most abundant of the IgG subclass
Human myeloma IgG1, λ is purified from human plasma by fractionation, ion-exchange, and affinity chromatography procedures.
IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. IgG is further subdivided into four subclass- IgG1, IgG2, IgG3 and IgG4.Elevation in antigen specific IgG1 is associated with symptomatic infection and Th2-regulation. IgG1 λ from human myeloma can be used in quantitative ELISA.Human IgG1 λ react specifically with human IgG.

Application

The purified IgG1, λ may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays. Human myeloma IgG1, λ was used as non-specific control in quantitative ELISA and solid-phase ELISA. A range of concentrations of IgG1, λ from 10 to 100 ng was used for quantitative ELISA. Human myeloma IgG1, λ was also used as control in size-exclusion chromatography to analyse purity of recombinant antibodies.
IgG1 λ from human myeloma can be used as reference antigen, blocking agent, immunoglobulin calibrator or coating protein in various immunoassay like ELISA, dot blot immunobinding, immunodiffusion, immunoelectrophoresis, hemagglutination, cell-binding assays and western immunoblotting.

Physical form

Solution in 0.02 M Tris buffered saline, pH 8.0.

Analysis Note

Identity is determined by immunoelectrophoresis and indirect ELISA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I5529M6034L0665
description

purified immunoglobulin, >95% (SDS-PAGE)

description

purified immunoglobulin, >90% (microfluidic capillary gel electrophoresis)

description

clone HOPC-1, purified immunoglobulin, buffered aqueous solution

description

purified immunoglobulin, ~1.0 mg/mL, ≥99% (Purity by double diffusion), buffered aqueous solution

biological source

human

biological source

human

biological source

mouse

biological source

human

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

assay

>95% (SDS-PAGE)

assay

>90% (microfluidic capillary gel electrophoresis)

assay

-

assay

≥99% (Purity by double diffusion)


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters