Skip to Content
Merck
All Photos(2)

Key Documents

HPA037837

Sigma-Aldrich

Anti-IPMK antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Inositol polyphosphate multikinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

DCFDGVLLELRKYLPKYYGIWSPPTAPNDLYLKLEDVTHKFNKPCIMDVKIGQKSYDPFASSEKIQQQVSKYPLMEEIGFLVLGMRVYHVHSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IPMK(253430)

General description

The gene encoding inositol polyphosphate multikinase (IPMK) enzyme is located on human chromosome 10q21.1. IPMK consists of inositol phosphate binding, nuclear localization signal, and ATP-binding kinase domain. It is a catalytically flexible enzyme and is part of intracellular signalling network.

Immunogen

inositol polyphosphate multikinase recombinant protein epitope signature tag (PrEST)

Application

Anti-IPMK antibody produced in rabbit may be used for the detection of IPMK protein in
  • lymphoblasts by immunoprecipitation
  • human adenocarcinogenic cell lines by western blotting
  • in mouse 3T3 cells by western blotting

Biochem/physiol Actions

Inositol polyphosphate multikinase (IPMK) indirectly mediates mRNA transport from nucleus to cytoplasm by mediating synthesis of phosphatidylinositol levels. IPMK interacts and stabilizes tumor necrosis factor receptor in HEK293T cells.IPMK coordinates with various signaling networks. Its deletion impairs immune response signalling pathways. Knockdown of IPMK leads to imbalance in the inositol polyphosphates. IPMK binds to tumor suppressor protein and regulates transcription and cell death. Mutation in the IPMK gene results in a truncated protein with reduced kinase activity. IPMK gene deletion or RNA interference results in cell growth inhibition. IPMK is regarded as prime target for tumor suppression. Pathology of Huntington′s disease is associated with IPMK protein depletion. In Alzheimer′s patients, low IPMK transcript levels may play a key in neurodegeneration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80294

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human homologue of yeast ArgRIII protein is an inositol phosphate multikinase with predominantly nuclear localization
Nalaskowski MM, et al.
The Biochemical Journal, 366(2), 549-556 (2002)
Radiosensitization of tumour cell lines by the polyphenol Gossypol results from depressed double-strand break repair and not from enhanced apoptosis
Kasten-Pisula U, et al.
Radiotherapy and Oncology : Journal of the European Society for Therapeutic Radiology and Oncology, 83(3), 296-303 (2007)
Association between genetic traits for immune-mediated diseases and Alzheimer disease
Yokoyama JS, et al.
JAMA Neurology, 73(6), 691-697 (2016)
Inositol polyphosphate multikinase promotes Toll-like receptor-induced inflammation by stabilizing TRAF6
Kim E, et al.
Science Advances, 3(4), e1602296-e1602296 (2017)
Inositol polyphosphate multikinase is a physiologic PI3-kinase that activates Akt/PKB
Maag D, et al.
Proceedings of the National Academy of Sciences of the USA, 108(4), 1391-1396 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service