Skip to Content
Merck
All Photos(1)

Key Documents

SAB5500134

Sigma-Aldrich

Anti-KI-67 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP6, tissue culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

SP6, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

bovine

technique(s)

immunohistochemistry: 1:200

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MKI67(4288)

General description

Ki-67 is a nuclear protein, which is expressed in the proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.
The gene marker of proliferation Ki-67 (MKI67), spanning 15 exons, is mapped to human chromosome 10q26.2. The encoded protein contains phosphopeptide-binding forkhead-associated (FHA) domain and a protein phosphatase 1 (PP1)-binding site at N- terminal. It possesses 16 tandem repeats at central region and leucine and arginine (LR) pairs at C- terminal end.

Immunogen

Synthetic peptide from C-terminus of human Ki-67 protein.

Application

Anti-KI-67 antibody, Rabbit monoclonal has been used in immunohistochemistry.

Biochem/physiol Actions

Marker of proliferation Ki-67 (MKI67), which is a part of the mitotic chromosome periphery, functions as a biological surfactant to maintain individual mitotic chromosomes dispersed in the cytoplasm after nuclear envelope disassembly. Ki-67 is a vital prognostic marker in breast and prostate cancer. Expression of the encoded protein is associated with cell-proliferation rate. Ki-67 is expressed at all active phases of the cell cycle (G (1), S, G (2), and mitosis), except resting cells (G (0)). Therefore, this protein is considered to be a potent cell proliferation marker.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody supplied as tissue culture supernatant in TBS/1% BSA buffer pH 7.5 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ki67 labelling index: an independent predictor of progression in prostate cancer treated by radical prostatectomy.
Bubendrof L, et al.
The Journal of Pathology, 178(4), 437-441 (1996)
Veronique L Veenstra et al.
Molecular oncology, 11(8), 1050-1064 (2017-05-10)
Pancreatic ductal adenocarcinoma (PDAC) is marked by an abundant stromal deposition. This stroma is suspected to harbor both tumor-promoting and tumor-suppressing properties. This is underscored by the disappointing results of stroma targeting in clinical studies. Given the complexity of tumor-stroma
The Ki?67 protein: from the known and the unknown.
Scholzen T and Johannes G
Journal of Cellular Physiology, 182(3), 311-322 (2000)
Ki-67 is a prognostic parameter in breast cancer patients: results of a large population-based cohort of a cancer registry.
Inwald E C, et al.
Breast Cancer Research and Treatment, 139(2), 539-552 (2013)
Job Komen et al.
Micromachines, 13(5) (2022-05-29)
The cancer xenograft model in which human cancer cells are implanted in a mouse is one of the most used preclinical models to test the efficacy of novel cancer drugs. However, the model is imperfect; animal models are ethically burdened

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service