Skip to Content
Merck
All Photos(4)

Key Documents

HPA029691

Sigma-Aldrich

Anti-SIRT4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-SIR2L4, Anti-sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
9
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VLQERFQVLNPTWSAEAHGLAPDGDVFLSEEQVRSFQVPTCVQCGGHLKPDVVFFGDTVNPDKVDFVHKRVKEADSLLVVGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT4(23409)

General description

SIRT4 (sirtuin 4) belongs to sirtuin family of nicotinamide adenine dinucleotide-dependent enzymes. It is a NAD+-dependent ADP ribosyltransferase, that is located in the mitochondria. SIRT4 protein level is found to be more in ESCC (esophageal squamous cell carcinoma) tissues.

Immunogen

sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae) recombinant protein epitope signature tag (PrEST)

Application

Anti-SIRT4 has been used in immunohistochemistry.

Biochem/physiol Actions

SIRT4 (sirtuin 4) may involve in the development of esophageal cancer. Growth of HeLa cells can be suppressed by the overexpression of SIRT4. It modulates glutamine metabolism and may also act as a tumor suppressor. It plays some important roles in multiple cellular processes like stress response and longevity. It may act as an important therapeutic target in colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78151

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumour-suppressive function of SIRT4 in human colorectal cancer
Miyo M, et al.
British Journal of Cancer, 113(3), 492-499 (2015)
Zhouxun Chen et al.
OncoTargets and therapy, 12, 2397-2408 (2019-04-18)
SIRT4, a protein localized in the mitochondria, is one of the least characteristic members of the sirtuin family. It is known that SIRT4 has deacetylase activity and plays a role in energy metabolism, but little is known about its possible
Yiwang Hu et al.
Oncology letters, 17(2), 2171-2176 (2019-02-13)
The sirtuins (SIRTs) are a family of nicotinamide-adenine dinucleotide (NAD)+-dependent protein deacetylases. SIRT4 is a mitochondrial NAD+-dependent adenosine diphsophate-ribosyltransferase. Recent studies demonstrated that SIRT4 can regulate glutamine metabolism and thus act as a tumor suppressor. However, the association of SIRT4
SIRT4 is upregulated in Chinese patients with esophageal cancer
Lai X, et al.
International Journal of Clinical and Experimental Pathology, 9(10), 10543-10549 (2016)
Frank K Huynh et al.
American journal of physiology. Endocrinology and metabolism, 319(4), E805-E813 (2020-09-01)
Sirtuins are a family of proteins that regulate biological processes such as cellular stress and aging by removing posttranslational modifications (PTMs). We recently identified several novel PTMs that can be removed by sirtuin 4 (SIRT4), which is found in mitochondria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service