UFSC05001
Amicon® Stirred Cell 50mL
This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.
Synonym(s):
Stirred Cell
About This Item
Recommended Products
material
316 stainless steel ring(s) (stir bar retaining)
316 stainless steel washer
Nylon connections (Luer on filtrate tubing)
Tygon® tubing (filtrate; E-3603)
polyacetal base
polyacetal support (for stir bar)
polyethylene tubing (pressure)
polysulfone body
polysulfone cap
polysulfone holder (for membrane)
polysulfone stir bar(s)
silicone O-ring
silicone gasket
Quality Level
product line
Amicon®
manufacturer/tradename
Amicon®
parameter
50 mL process volume
technique(s)
protein clean-up: suitable (desalting)
protein purification: suitable (concentration)
H
11.7 cm (4.6 in.)
filter diam.
44.5 mm
filtration area
13.4 cm2
shipped in
ambient
Application
Principle
Linkage
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.
Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.
Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.
Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service