Skip to Content
Merck
All Photos(10)

Key Documents

HPA019527

Sigma-Aldrich

Anti-AMACR antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-methylacyl-CoA racemase, Anti-RACE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:2500-1:5000
western blot: 0.04-0.4 μg/mL

immunogen sequence

HNKERGSFITSEEQDVSPRPAPLLLNTPAIPSFKRDPFIGEHTEEILEEFGFSREEIYQLNSDKIIESNKVKASL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMACR(23600)

General description

AMACR (a-methylacyl-CoA racemase) is a 43,304 daltons auxiliary racemase protein expressed in peroxisomes and mitochondria of human liver and fibroblasts. It consists of 382 amino acids, two targeting signals, a mitochondrial sequence at its N terminus, and a peroxisomal sequence at its C terminus.

Immunogen

alpha-methylacyl-CoA racemase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AMACR (a-methylacyl-CoA racemase) plays a crucial role in the fatty acid metabolism. It mediates the peroxisomal ß-oxidative breakdown of (2R)-pristanic acid and the (25R)-isomer of C(27) bile acid intermediates. Mutation in AMACR gene causes adult-onset sensory motor neuropathy. It causes unnecessary accumulation of specific fatty acids.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74752

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Amery et al.
Journal of lipid research, 41(11), 1752-1759 (2000-11-04)
2-Methylacyl-CoA racemase is an auxiliary enzyme required for the peroxisomal beta-oxidative breakdown of (2R)-pristanic acid and the (25R)-isomer of C(27) bile acid intermediates. The enzyme activity is found not only in peroxisomes but also is present in mitochondria of human
Yachana Kataria et al.
The Prostate, 75(2), 200-210 (2014-10-14)
Alpha-methylacyl-CoA racemase (AMACR) is an enzyme involved in fatty acid metabolism that is markedly over-expressed in virtually all prostate cancers (PCa), relative to benign tissue. One of AMACR's primary substrates, phytanic acid, is derived predominately from red meat and dairy
Jiwei Yao et al.
ACS omega, 3(6), 6411-6418 (2018-07-20)
Prostate cancer is prevalent among cancers in men. A simple method for screening of reliable biomarkers is pivotal for early detection of prostate cancer.  Prostate-specific antigen (PSA) has been a commonly used biomarker for prostate cancer, in spite of its
S Ferdinandusse et al.
Nature genetics, 24(2), 188-191 (2000-02-02)
Sensory motor neuropathy is associated with various inherited disorders including Charcot-Marie-Tooth disease, X-linked adrenoleukodystrophy/adrenomyeloneuropathy and Refsum disease. In the latter two, the neuropathy is thought to result from the accumulation of specific fatty acids. We describe here three patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service