Skip to Content
Merck
All Photos(3)

Key Documents

AB5078P

Sigma-Aldrich

Anti-Beta (β)-Amyloid antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Anti-AAA, Anti-ABETA, Anti-ABPP, Anti-AD1, Anti-APPI, Anti-CTFgamma, Anti-CVAP, Anti-PN-II, Anti-PN2, Anti-alpha-sAPP, Anti-preA4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Beta-Amyloid 1-42 Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

chicken, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

The cerebral and vascular plaques associated with Alzheimer′s disease (AD) are mainly composed of amyloid beta peptides (Ab). Ab is derived from cleavage of the amyloid precursor protein (APP) and varies in length from 39 to 43 amino acids. Ab [1-40], Ab [1-42], and Ab [1-43] peptides result from cleavage of APP after residues 40, 42, and 43, respectively. The cleavage takes place by gamma-secretase during the last APP processing step. Ab [1-40], [1-42] and [1-43] peptides are major constituents of the plaques and tangles that occur in AD. Ab antibodies and peptides have been developed as tools for elucidating the biology of AD.

Specificity

Recognizes beta-amyloid 1-42. Antibody does not show appreciable reactivity with beta amyloid 1-40 as detected by ELISA.
One of the most important and initial steps which causes loss of memory and cognition in Alzheimer’s Disease (AD) involves proteolytic cleavage of amyloid precursor protein (APP, chromosome 21) releasing short 40, 42 & 43 amino acid peptides (beta amyloid 1-40, 1-42 and 1-43). Polymerization of beta-amyloid and subsequent neuronal deposit (amyloid) leads to the degeneration of neurons involved in memory and cognition.

Immunogen

A six amino acid peptide sequence from the C-terminal of human beta-amyloid 1-42.
Epitope: C-terminus

Application

Anti-Beta-Amyloid 1-42 Antibody is an antibody against Beta-Amyloid 1-42 for use in ELISA, IH, IH(P) & WB.
Immunohistochemistry (Paraffin) Analysis:

Western Blot:
1-10 µg/mL of a previous lot was used. (Chemiluminescence technique).

ELISA:
1:10,000-1:100,000 of a previous lot was used using 50-100 ng immunogen peptide/well.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Routinely evaluated by immunohistochemistry (paraffin) on Alzheimer’s Disease-Brain.

Immunohistochemistry (Paraffin) Analysis:
Beta-Amyloid (cat. # AB5078P) staining on Alzheimer’s Disease-Brain. Tissue pretreated with Citrate, pH 6.0. The antibody was diluted to 1:35, using IHC-Select Detection with HRP-DAB. Immunoreactivity is seen as staining on plaque deposits (brown) or as cerebral vascular amyloid deposition in the walls of blood vessels (brown).

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal in liquid buffer containing PBS with 0.1% BSA.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Alzheimer′s disease brain tissue, whole tissue extracts from mouse brain. C03-801.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic thrombin exposure results in an increase in apolipoprotein-E levels.
Mhatre, M; Hensley, K; Nguyen, A; Grammas, P
Journal of Neuroscience Research null
Accumulation of intraneuronal ?-amyloid 42 peptides is associated with early changes in microtubule-associated protein 2 in neurites and synapses.
Takahashi, RH; Capetillo-Zarate, E; Lin, MT; Milner, TA; Gouras, GK
Testing null
Athanassios Fragoulis et al.
Redox biology, 12, 843-853 (2017-04-28)
There is increasing evidence for the involvement of chronic inflammation and oxidative stress in the pathogenesis of Alzheimer's disease (AD). Nuclear factor erythroid 2-related factor 2 (Nrf2) is an anti-inflammatory transcription factor that regulates the oxidative stress defense. Our previous
Ramon Velazquez et al.
Molecular neurodegeneration, 11(1), 52-52 (2016-07-15)
Alzheimer's disease (AD) is the most prevalent neurodegenerative disorder worldwide. Clinically, AD is characterized by impairments of memory and cognitive functions. Accumulation of amyloid-β (Aβ) and neurofibrillary tangles are the prominent neuropathologies in patients with AD. Strong evidence indicates that
Constantinos D Paspalas et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 14(5), 680-691 (2017-12-16)
An animal model of late-onset Alzheimer's disease is needed to research what causes degeneration in the absence of dominant genetic insults and why the association cortex is particularly vulnerable to degeneration. We studied the progression of tau and amyloid cortical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service