Skip to Content
Merck
All Photos(1)

Key Documents

H9384

Sigma-Aldrich

22(R)-Hydroxycholesterol

≥98%

Synonym(s):

22α-Hydroxycholesterol, 5-Cholestene-3β,22(R)-diol, 5-Cholestene-3β,22[R]-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC=C3C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])[C@H](C)[C@H](O)CCC(C)C

InChI

1S/C27H46O2/c1-17(2)6-11-25(29)18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20-,21-,22+,23-,24-,25+,26-,27+/m0/s1

InChI key

RZPAXNJLEKLXNO-GFKLAVDKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bovine aortic endothelial cells were treated with 22(R)-Hydroxycholesterol to study the effects on production of free radicals and in studies related to fatty acid metabolism.

Biochem/physiol Actions

22(R)-Hydroxycholesterol is an intermediate of the pregnenolone synthesis pathway from cholesterol. It reported has neuroprotective properties and protects the neurons against β-amyloid-induced cell death. 22(R)-Hydroxycholesterol acts as the ligand of liver X receptors that act as sensors of sterol concentration and regulates the fatty acid metabolism.

Preparation Note

22(R)-Hydroxycholesterol yield clear, colorless solution in chloroform.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Chawla et al.
Science (New York, N.Y.), 294(5548), 1866-1870 (2001-12-01)
Cholesterol, fatty acids, fat-soluble vitamins, and other lipids present in our diets are not only nutritionally important but serve as precursors for ligands that bind to receptors in the nucleus. To become biologically active, these lipids must first be absorbed
Cynthia Hong et al.
Journal of lipid research, 52(3), 531-539 (2010-12-29)
Ligand activation of liver X receptors (LXRs) has been shown to impact both lipid metabolism and inflammation. One complicating factor in studies utilizing synthetic LXR agonists is the potential for pharmacologic and receptor-independent effects. Here, we describe an LXR gain-of-function
Lourdes Cruz-Garcia et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 160(2), 125-136 (2011-06-04)
The liver X receptor (LXR) has recently been described in salmonids. In mammals, this receptor is already known as a transcriptional factor that regulates diverse aspects of cholesterol, fatty acid and carbohydrate metabolism in various tissues, including muscle. Here we
Linara Gabitova et al.
Cell reports, 12(11), 1927-1938 (2015-09-08)
Meiosis-activating sterols (MAS) are substrates of SC4MOL and NSDHL in the cholesterol pathway and are important for normal organismal development. Oncogenic transformation by epidermal growth factor receptor (EGFR) or RAS increases the demand for cholesterol, suggesting a possibility for metabolic
Nina Hallmark et al.
Environmental health perspectives, 115(3), 390-396 (2007-04-14)
Certain phthalates can impair Leydig cell distribution and steroidogenesis in the fetal rat in utero, but it is unknown whether similar effects might occur in the human. Our aim in this study was to investigate the effects of di(n-butyl) phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service