Skip to Content
Merck
All Photos(1)

Key Documents

H6378

Supelco

20α-Hydroxycholesterol

analytical standard

Synonym(s):

5-Cholestene-3β,20α-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC(C)CCC[C@](C)(O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O2/c1-18(2)7-6-14-27(5,29)24-11-10-22-21-9-8-19-17-20(28)12-15-25(19,3)23(21)13-16-26(22,24)4/h8,18,20-24,28-29H,6-7,9-17H2,1-5H3/t20-,21-,22-,23-,24-,25-,26-,27-/m0/s1

InChI key

MCKLJFJEQRYRQT-APGJSSKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

20α-Hydroxycholesterol has been used as a standard to determine the concentration of 20α-hydroxycholesterol, present in food samples using gas chromatography with an ion trap detector coupled with mass spectrometer (GC-IT-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

B C McNamara et al.
Molecular and cellular endocrinology, 73(2-3), 123-134 (1990-10-22)
Side-chain cleavage (SCC) of endogenous cholesterol in adrenal mitochondria isolated from ACTH-treated rats indicates that the size of the reactive cholesterol pool depends on the reducing precursor. At optimal concentrations of reductant, this pool was typically at least 2 times
Ying Yong Lin et al.
The Journal of steroid biochemistry and molecular biology, 85(1), 57-61 (2003-06-12)
The presence of 20(S)-hydroxycholesterol in rat brains and human placenta has been established using the gas chromatography/mass spectrometry (GC/MS) select ion monitoring (SIM) technique. Identification was ensured by three criteria: the specific retention time when the compound emerges from the
J W Laskey et al.
Toxicology and applied pharmacology, 108(2), 296-306 (1991-04-01)
In vivo assessment of toxicant action on Leydig cell function is subject to homeostatic mechanisms which make it difficult to determine whether any changes seen in serum testosterone (T) concentration are due to extragonadal endocrine alterations or to a direct
K Z Liu et al.
American heart journal, 123(2), 285-290 (1992-02-01)
Oxidized low-density lipoprotein (LDL) may play an important role in atherogenesis. The oxidative status of isolated LDL and very low-density lipoprotein (VLDL) were investigated in 23 patients with proven coronary disease and in 23 healthy asymptomatic control subjects. Oxidized cholesterol
H Guo et al.
Endocrinology, 127(3), 1234-1239 (1990-09-01)
Interstitial tissue of the testis consists of Leydig cells, macrophages, lymphocytes, plasma cells, mast cells and fibroblasts. Previously we have reported that interleukin-1 (IL-1) inhibits Leydig cell androgen production. In the present study, the effect of IL-2 was investigated. Leydig

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service