H5134
D-Homocystine
≥98% (TLC)
Synonym(s):
(R,R)-4,4′-Dithiobis(2-aminobutanoic acid)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H16N2O4S2
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
D-Homocystine,
Assay
≥98% (TLC)
Quality Level
form
powder
color
white to off-white
SMILES string
N[C@H](CCSSCC[C@@H](N)C(O)=O)C(O)=O
InChI
1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6-/m1/s1
InChI key
ZTVZLYBCZNMWCF-PHDIDXHHSA-N
Biochem/physiol Actions
D-Homocystine may be used in the preparation of carbocyclic nucleoside such as S-aristeromycinyl-L-homocysteine for use in studied on S-adenosylmethionine-dependent methyltransferase inhibitors.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
D M Houston et al.
Journal of medicinal chemistry, 28(4), 478-482 (1985-04-01)
A series of base- and amino acid modified analogues of S-aristeromycinyl-L-homocysteine, a carbocyclic nucleoside, were synthesized and evaluated as inhibitors of S-adenosyl-L-methionine-dependent methyltransferases, including catechol O-methyltransferase, phenylethanolamine N-methyltransferase, and histamine N-methyltransferase. The base-modified analogues (8-azaadenine, 3-deazaadenine, and N6-methyladenine) were prepared
S-Aristeromycinyl-L-homocysteine, a potent inhibitor of S-adenosylmethionine-dependent transmethylations.
R T Borchardt et al.
Journal of medicinal chemistry, 19(1), 197-198 (1976-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service