Skip to Content
Merck
All Photos(3)

Key Documents

C7505

Sigma-Aldrich

L-Cystathionine

≥98% (TLC), suitable for UHPLC

Synonym(s):

(R)-S-(2-Amino-2-carboxyethyl)-L-homocysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
Beilstein:
2505200
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Cystathionine, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

UHPLC: suitable

color

white

mp

>300 °C

storage temp.

−20°C

SMILES string

N[C@@H](CCSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m0/s1

InChI key

ILRYLPWNYFXEMH-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cystathionine has been used in ultra-performance liquid chromatography for the validation of cystathionine and the correlation of its abundance with vertebrate metamorphosis. It has also been used as an inhibitor of sinusoidal GSH (glutathione) carrier to signify that thiol redox state imbalance is associated with the modulation of autophagy in carcinoma cells.[1]

Biochem/physiol Actions

L-Cystathionine is an intermediate in the biosynthesis of L-cysteine and methionine.[2][3] It is used as a substrate to differentiate and analyze cystathionine γ-lyase(s).[4] L-Cystathionine is an important non-protein amino acid and is associated with metabolic disorders.[5] In recent studies, L-cystathionine is believed to be the precursor of cyclic imino acid cyclothionine. Cystathioninuria is characterized with imino acid cyclothionine excretion in the urine.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The transamination of L-cystathionine, L-cystine and related compounds by a bovine kidney transaminase.
Ricci G
European Journal of Biochemistry, 157(1), 57-63 (1986)
Amany K Elshorbagy et al.
Nutrition (Burbank, Los Angeles County, Calif.), 26(11-12), 1201-1204 (2010-01-19)
Dietary methionine restriction in Fischer-344 rats favorably influences visceral fat mass, insulin sensitivity, metabolic parameters, and longevity. However, little is known about the effects of methionine restriction on serum methionine and its downstream sulfur amino acids. We investigated the serum
Synthesis and evaluation of L-cystathionine as a standard for amino acid analysis.
Amino Y
Bioscience, Biotechnology, and Biochemistry, 81(1), 95-101 (2017)
Zoltan Dekan et al.
Journal of the American Chemical Society, 133(40), 15866-15869 (2011-09-09)
The two disulfide bonds of α-conotoxin ImI, a peptide antagonist of the α7 nicotinic acetylcholine receptor (nAChR), were systematically replaced with isosteric redox-stable cystathionine thioethers. Regioselective thioether formation was accomplished on solid support through substitution of a γ-chlorohomoalanine by an
Amany K Elshorbagy et al.
Current opinion in clinical nutrition and metabolic care, 15(1), 49-57 (2011-11-24)
The concentrations of several plasma amino acids increase in obesity. Notably, plasma total concentrations of the sulphur amino acid cysteine (tCys) are linearly associated with fat mass in large population studies. Animal and cellular experiments support the concept that cysteine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service