Skip to Content
Merck
All Photos(2)

Key Documents

H2625

Sigma-Aldrich

Hemoglobin from bovine blood

suitable for protease substrate, substrate powder

Synonym(s):

Bovine hemoglobin, Hb, Methemoglobin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42030116
NACRES:
NA.61

biological source

bovine blood

form

substrate powder

mol wt

Mr ~64500

technique(s)

activity assay: suitable

solubility

H2O: soluble 20 mg/mL

suitability

suitable for protease substrate

UniProt accession no.

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Hemoglobin is the major component of red blood cells and is responsible for their red color. Its normal concentration in erythrocytes is 34%. Hemoglobin is a globular protein with α and β chains with every 141 and 146 amino acids respectively. It exists as a tetramer with each monomer having heterocyclic porphyrin ring with iron constituting the heme.

Application

Hemoglobin from bovine blood has been used as a substrate in cathepsin D activity assay. It has also been used to measure the activity of acid proteases (pepsin-like) in the stomach extract.
The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.

Biochem/physiol Actions

Bovine hemoglobin is used in the production of hemoglobin-vesicles (HbV). It has high thermal stability and high oxygen affinity when compared to human hemoglobin. Bovine hemoglobin interacts with synthetic and azo dyes. Polymerized forms of bovine hemoglobin are used to treat autoimmune hemolytic anemia.
Oxygen transporter, NO scavenger
Oxygen transporter. The Fe2+/Fe3+ balance is a physiological indicator of blood oxygenation; deoxygenated hemoglobin accessorizes a feedback loop by reducing nitrite to NO, a vasodilator which enhances blood flow to oxygen-deprived tissues.

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Preparation Note

Prepared from washed, lysed and dialyzed erythrocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High pressure effects on the activities of cathepsins B and D of mackerel and horse mackerel muscle
Fidalgo L, et al.
Czech Journal of Food Sciences, 32(2), 188-193 (2016)
P John Wright et al.
Journal of the American Society for Mass Spectrometry, 20(3), 484-495 (2008-12-23)
The conformations of gas-phase ions of hemoglobin, and its dimer and monomer subunits have been studied with H/D exchange and cross section measurements. During the H/D exchange measurements, tetramers undergo slow dissociation to dimers, and dimers to monomers, but this
Circadian cycle of digestive enzyme production at fasting and feeding conditions in Nile tilapia, Oreochromis niloticus (Actinopterygii: Perciformes: Cichlidae)
MEJIA, MAGNOLIA MONTOYA and others
Acta ichthyologica et piscatoria, 32(2), 188-193 (2016)
Characteristics of bovine hemoglobin as a potential source of hemoglobin-vesicles for an artificial oxygen carrier
Sakai H, et al.
Journal of Biochemistry, 131(4), 611-617 (2002)
Luke J Norbury et al.
Biochimie, 93(3), 604-611 (2010-12-21)
Cathepsin proteases are promising vaccine or drug targets for prophylaxis or therapy against Fasciola parasites which express cathepsin L and B proteases during their development. These proteases are believed to be involved in important functions for the parasite, including excystment

Protocols

This procedure may be used for determination of Pepsin activity using hemoglobin as the substrate. It is a spectrophotometric stop rate determination.

Proteinase K activity measured via spectrophotometry using hemoglobin substrate, crucial for enzyme characterization.

Proteinase K activity measured via spectrophotometry using hemoglobin substrate, crucial for enzyme characterization.

Proteinase K activity measured via spectrophotometry using hemoglobin substrate, crucial for enzyme characterization.

See All

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service