Skip to Content
Merck
All Photos(2)

Documents

08449

Millipore

Hemoglobin from bovine blood

suitable for microbiology

Synonym(s):

FO Growth Supplement, Bovine hemoglobin, Hb, Methemoglobin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41171614
eCl@ss:
42030116
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

mol wt

Mr ~64500

loss

<10% loss on drying

UniProt accession no.

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Neisseria spp.

Looking for similar products? Visit Product Comparison Guide

General description

Hemoglobin is present in the red-blood corpuscles (RBCs) of vertebrates. It is composed of a globular structure with four α-β sub-units.

Application

Hemoglobin from bovine blood has been used as a supplement in tryptone yeast extract salts and glucose (TYESG) agar for performing extracellular proteolytic activity of Flavobacterium psychrophilum. It has also been used as a standard in Raman spectrometry.
The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.

Biochem/physiol Actions

Oxygen transporter. The Fe2+/Fe3+ balance is a physiological indicator of blood oxygenation; deoxygenated hemoglobin accessorizes a feedback loop by reducing nitrite to NO, a vasodilator which enhances blood flow to oxygen-deprived tissues.

Packaging

1 piece contains 100 g hemoglobin powder

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamaljeet et al.
Frontiers in chemistry, 4, 50-50 (2017-01-26)
Synthetic dyes are a very efficient class of dyes that are ingested or come into contact with the skin from numerous sources (cosmetics, textiles, leather, paper, and drugs). An important component of their safety profile is the interactions that they
Jennie B Leach et al.
Acta biomaterialia, 1(2), 155-164 (2006-05-17)
Elastin is a critical biochemical and biomechanical component of vascular tissue. However, elastin is also highly insoluble and therefore difficult to process into new biomaterials. We present a simple approach for synthesizing elastin-based materials from two commercially available and water-soluble
K M Marzec et al.
The Analyst, 140(7), 2171-2177 (2014-12-24)
The composition of the lung tissue of mice was investigated using Raman confocal microscopy at 532 nm excitation wavelength and was supported with various staining techniques as well as DFT calculations. This combination of experimental and theoretical techniques allows for
Paul Barbier et al.
Applied and environmental microbiology, 86(16) (2020-06-14)
Flavobacterium psychrophilum causes bacterial cold-water disease in wild and aquaculture-reared fish and is a major problem for salmonid aquaculture. The mechanisms responsible for cold-water disease are not known. It was recently demonstrated that the related fish pathogen, Flavobacterium columnare, requires
N F Schreiter et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 185(4), 358-366 (2013-03-16)
To investigate whether changes in hemodynamics induced by Valsalva maneuver can be exploited for detecting and characterizing breast lesions by optical mammography. 30 women underwent optical imaging of the breast using a DYNOT 232 system and performing Valsalva maneuvers prior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service