GE29-0485-86
HisTrap™ Excel
Cytiva 29-0485-86, pack of 1 mL
About This Item
Recommended Products
packaging
pack of 1 mL
manufacturer/tradename
Cytiva 29-0485-86
availability
not available in North America
parameter
1 ml/min - 4 ml/min flow rate
5 bar
bed size
7 mm × 25 mm
bed volume
1 mL
column I.D.
7 mm
matrix
highly cross-linked 6% agarose
average diameter
90 μm
cleaning
2-14
working range
2-13
General description
Features and Benefits
- Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity
- Increase target protein yield and decrease degradation through reduced and simplified sample handling
- HisTrap excel columns allow direct purification of cell-free, unclarified samples
- HisTrap excel columns are made of biocompatible polypropylene. The columns are delivered with a stopper on the inlet and a snap-off end on the outlet.
- HisTrap excel columns can be operated either with a peristaltic pump or with chromatography systems such as ÄKTA systems. The columns cannot be opened or refilled.
Storage and Stability
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.
Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.
Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.
Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.
Protocols
This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using HisTrap Excel from Cytiva.
How to isolate proteins from inclusion bodies using Cytiva products.
This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service