Skip to Content
Merck

70666

Ni-NTA His•Bind® Resin

Synonym(s):

nickel affinity gel, nickel charged agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41106500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

slurry

Quality Segment

manufacturer/tradename

Novagen®

storage condition

do not freeze

application(s)

life science and biopharma

shipped in

ambient

storage temp.

2-8°C

General description

Ni-NTA His•Bind Resin is a high-performance Ni2+-charged agarose used for rapid one-step purification of proteins containing a His•Tag sequence by metal chelation chromatography. NTA chemistry minimizes metal leaching during purification and is compatible with up to 10 mM β-mercaptoethanol or 1 mM Tris(hydroxypropyl)phosphine (THP) for reduction of disulfide bonds. Ni-NTA His•Bind Resin has a binding capacity of 5–10 mg His•Tag fusion protein per ml resin.

Supplied as a 50% slurry; the quantity/pack is based on the amount of the settled resin.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

HIS-BIND is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Flammable (J)


pictograms

Flame

Storage Class

3 - Flammable liquids

flash_point_f

86.0 °F

flash_point_c

30 °C

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hadas Bar-Joseph et al.
Reproductive biomedicine online, 41(2), 335-342 (2020-06-14)
Does recombinant pigment epithelium derived factor (PEDF) have potential in treating uterine fibroids? In-vitro models that used human leiomyoma and Eker rat uterine leiomyoma (ELT-3) cell lines. The ELT-3 cell line was used to examine cellular targets after adding recombinant
Yu-Xi Luo et al.
The EMBO journal, 39(7), e102008-e102008 (2020-03-03)
Deposition of H2A.Z in chromatin is known to be mediated by a conserved SWR1 chromatin-remodeling complex in eukaryotes. However, little is known about whether and how the SWR1 complex cooperates with other chromatin regulators. Using immunoprecipitation followed by mass spectrometry
Bao-Rui Zhao et al.
Journal of immunology (Baltimore, Md. : 1950), 204(3), 487-497 (2019-12-20)
Rapid synthesis and release of active antimicrobial peptides (AMPs) is an important strategy in innate immune. Processing of the precursor into the active form is a common posttranslational modification of AMPs in mammals. However, in invertebrates, the mechanism of AMP



Global Trade Item Number

SKUGTIN
70666-307790788054946
70666-604055977256826
70666-407790788052768
70666-504055977334074