Skip to Content
Merck
All Photos(1)

Key Documents

GE27-0798-04

Recombinant Taq DNA Polymerase

Cytiva 27079804

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.55

recombinant

expressed in E. coli

form

liquid

manufacturer/tradename

Cytiva 27079804

concentration

5000 unit/mL

color

colorless

pH

7.5

solubility

soluble (Soluble in cold water and hot water)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Taq DNA Polymerase (cloned) is the recombinant form of the native enzyme from Thermus aquaticus expressed in E. coli. Like native Taq, it polymerizes DNA from a primer annealed to a DNA template in the presence of deoxyribonucleotide triphosphates. Because Taq DNA Polymerase (cloned) is a recombinant protein, it is exceptionally pure and provides excellent batch-to-batch reproducibility. It has an optimum temperature of 75°C and can survive repeated incubations at > 95°C

Application

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

Features and Benefits

Highly purified thermostable DNA polymerase for PCR.

  • Highly purified recombinant enzyme.
  • Excellent batch-to-batch reproducibility.
  • Each lot is tested to be free of nuclease contamination.

Packaging

100 mM Tris-HCl, pH 9.0, 15 mM MgCl2 and 500 mM KCl. Also supplied with 25 mM MgCl2 solution

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Forest City, California 94404, USA.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service