Skip to Content
Merck
All Photos(2)

Documents

GE17-0110-01

Sepharose 6B

Cytiva 17-0110-01, pack of 1 L

Synonym(s):

Agarose, Beaded

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

feature

autoclavable Autoclavable, 20 min at 120°C in pH 7

packaging

pack of 1 L

manufacturer/tradename

Cytiva 17-0110-01

matrix

6% agarose

particle size

45-165 μm

cleaning in place

4-9

working range

4-9

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@@H](O3)O[C@H]4[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]5[C@@H]6CO[C@H]5[C@H](O)[C@H](O)O6)[C@@H]4O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sepharose 6B is a well-proven agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

Sepharose is a bead-formed agarose-based gel filtration matrix. Sepharose is available with 3 different agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight.

Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents.

Features and Benefits

  • 6% Agarose gel filtration media.
  • Proven base matrix for coupling affinity ligands

Other Notes

Beaded agarose for fractionating molecules of high molecular weight. Cross-linked beaded agarose is more resistant to denaturing conditions, and thus offers more versatility in the choice of sample buffer and eluent. The approximate % agarose concentration is indicated by the product name (Ultrogel A%).

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Tomita et al.
Ultrasonics, 55, 1-5 (2014-08-19)
Sonoporation has the potential to deliver extraneous molecules into a target tissue non-invasively. There have been numerous investigations of cell membrane permeabilization induced by microbubbles, but very few studies have been carried out to investigate sonoporation by inertial cavitation, especially
Yan Hong et al.
Journal of chromatography. A, 1342, 30-36 (2014-04-02)
Previously, we studied bovine serum albumin (BSA) uptake to poly(ethylenimine) (PEI)-grafted Sepharose resins, and an ionic capacity (IC) range (600-740mmol/L) for steep increases of both protein capacity (qm) and effective pore diffusion coefficient (De) was found. In this work, seven
The agarose migration inhibition technique for in vitro demonstration of cell-mediated immunity in man. A review.
J E Clausen
Danish medical bulletin, 22(5), 181-194 (1975-07-01)
Sieving by agarose gels and its use during pulsed-field electrophoresis.
P Serwer
Biotechnology & genetic engineering reviews, 8, 319-343 (1990-01-01)
Andrea Trochine et al.
Experimental parasitology, 140, 33-38 (2014-03-19)
Benznidazole (Bzn) is a nitroimidazole drug currently used as first line treatment against Chagas disease, a neglected tropical disease caused by the flagellated protozoan Trypanosoma cruzi. Although the drug has been used since the late 1960s, its mechanism of action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service