Key Documents
GE10600057
Amersham Hybond® P Western blotting membranes, PVDF
pore size 0.2 μm, sheet W × L 200 mm × 200 mm, pkg of 25 ea
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva 10600057
sheet W × L
200 mm × 200 mm
pore size
0.2 μm pore size
General description
PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
This page describes troubleshooting strategies in the detection of GST-tagged proteins.
Protocols
This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.