Skip to Content
Merck
All Photos(1)

Key Documents

GE10600023

Amersham Hybond® P Western blotting membranes, PVDF

pore size 0.45 μm, roll W × L 300 mm × 4 m, pkg of 1 ea

Synonym(s):

Amersham PVDF membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600023

roll W × L

300 mm × 4 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.

Features and Benefits

  • For use with standard colorimetric and chemiluminescent detection methods for proteins of > Mr 20 000
  • Offers high physical strength for easy signal detection
  • Provides high protein retention even after harsh washing steps
  • Captures high protein during transfer reduces the loss of sample
  • Available in precut sizes for laboratory uses

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ousamah Younoss Soliman et al.
Journal of pharmaceutical sciences, 109(4), 1581-1593 (2020-01-01)
Messenger RNA (mRNA)-containing nanoparticles were produced by electrostatic complexation with a library of pharmaceutical grade chitosans with different degrees of deacetylation and hyaluronic acids (HAs) (native vs. sulfated). Polymer length (Mn), HA degree of sulfation (DS), and amine to phosphate
Jedediah Seltzer et al.
Journal of virology, 94(10) (2020-03-13)
Kaposi sarcoma-associated herpesvirus (KSHV) is necessary but not sufficient for primary effusion lymphoma (PEL) development. Alterations in cellular signaling pathways are also a characteristic of PEL. Other B cell lymphomas have acquired an oncogenic mutation in the myeloid differentiation primary
Narjes Khatoun Shabani Sadr et al.
Biochemical genetics, 58(6), 883-900 (2020-07-02)
Sialic acid (N-acetylneuraminic acid, NANA) is found at all cell surfaces of vertebrates. Although it is widely accepted that sialic acid is an essential substrate for brain development via a significant role in nerve transfers, structure of glycosides, and synaptogenesis
Massimo Venditti et al.
Animals : an open access journal from MDPI, 11(1) (2021-01-14)
Herein is reported the first evidence of the protective role of D-aspartic acid (D-Asp) in preventing the toxic effect exerted by the alkylating agent ethane dimethane sulfonate (EDS) in the rat testis. We confirmed that EDS treatment specifically destroyed Leydig
Lisa Mayr et al.
Nature communications, 11(1), 1775-1775 (2020-04-15)
The increased incidence of inflammatory bowel disease (IBD) has become a global phenomenon that could be related to adoption of a Western life-style. Westernization of dietary habits is partly characterized by enrichment with the ω-6 polyunsaturated fatty acid (PUFA) arachidonic

Articles

This page describes troubleshooting strategies in the detection of GST-tagged proteins.

Protocols

This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service