Skip to Content
Merck
All Photos(1)

Key Documents

G7795

Sigma-Aldrich

Ganaxolone

≥98% (HPLC), solid

Synonym(s):

3α-Hydroxy-3β-methyl-5α-pregnan-20-one, CCD 1042

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ≥2 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(C)=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@](C)(O)C2

InChI

1S/C22H36O2/c1-14(23)17-7-8-18-16-6-5-15-13-20(2,24)11-12-21(15,3)19(16)9-10-22(17,18)4/h15-19,24H,5-13H2,1-4H3/t15-,16-,17+,18-,19-,20+,21-,22+/m0/s1

InChI key

PGTVWKLGGCQMBR-FLBATMFCSA-N

Biochem/physiol Actions

Ganaxolone (3alpha-hydroxy-3beta-methyl-5alpha-pregnane-20-one) is a positive allosteric modulator of the GABAA receptor subtype; synthetic analog of the endogenous neurosteroid allopregnanolone; effective against chemically induced seizures in rats and mice. Ganaxolone is an orally active analog of allopregnanolone that is not converted to the hormonally active 3-keto form. The enhanced anticonvulsant potency of ganaxolone after neurosteroid withdrawal supports the use of ganaxolone as a specific treatment for perimenstrual catamenial epilepsy.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Quality

The product is pure based on elemental analysis, NMR, MS, optical rotation and melting point.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doodipala Samba Reddy
Progress in brain research, 186, 113-137 (2010-11-26)
This chapter provides an overview of neurosteroids, especially their impact on the brain, sex differences and their therapeutic potentials. Neurosteroids are synthesized within the brain and rapidly modulate neuronal excitability. They are classified as pregnane neurosteroids, such as allopregnanolone and
Vincent A Pieribone et al.
Epilepsia, 48(10), 1870-1874 (2007-07-20)
A pilot study of the safety, tolerability, dose range and potential efficacy of ganaxolone for the treatment of refractory epilepsy in pediatric and adolescent subjects. We report the results of a nonrandomized, nonblinded, open-label, dose-escalation trial of ganaxolone in pediatric
S H Hsiao et al.
Brain research. Developmental brain research, 130(1), 25-40 (2001-09-15)
Previously we found postnatal binge-like ethanol exposure using an artificial-rearing method in the rat delayed developmental up-regulation of GABA(A) receptors (GABA(A)Rs) in both medial septum/diagonal band (MS/DB) and cerebellar Purkinje neurons. In the present study, the impact of ethanol on
Marcia J Ramaker et al.
Neuropharmacology, 63(4), 555-564 (2012-05-23)
Recent evidence suggests that GABA(A) receptor ligands may regulate ethanol intake via effects at both synaptic and extrasynaptic receptors. For example, the endogenous neurosteroid, allopregnanolone (ALLO) has a similar pharmacological profile as ethanol, and it alters ethanol intake in rodent
Synthia H Mellon et al.
Brain research reviews, 57(2), 410-420 (2007-07-17)
The functions for neurosteroids during development and in response to nervous system injury are beginning to be identified. We focused on a mouse model in which we believed neurosteroid production would be altered, and which had a neurodegenerative phenotype. Niemann-Pick

Articles

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service